Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Available on GSA/VA Contract for Federal Government customers only. Corning™cellgro™ DMEM and Ham's F-12 50/50 Mix (Mod.)
Make only the quantity of media you need, when you need it. Media is formulated without sodium bicarbonate to prevent clumping due to moisture absorption. Supplied in plastic bottles. Shipped at ambient temperature; recommended storage temperature: 2° to 8°C (36° to 46°F).
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Corning™cellgro™ DMEM and Ham's F-12 50/50 Mix (Mod.)
(2 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
MT90090PB Available on GSA/VA Contract for Federal Government customers only. 
Mediatech
No.:90-090-PB
Size: 10L; Description: Without L-Glutamine, Phenol Red, and Sodium Bicarbonate
Each for $58.30
Check Availability
Add
to Cart
(0) in My Cart
MT90091PB Available on GSA/VA Contract for Federal Government customers only. 
Mediatech
No.:90-091-PB
Size: 10L; Description: Without L-Glutamine and Sodium Bicarbonate
Each for $67.10
Check Availability
Add
to Cart
(0) in My Cart
Make only the quantity of media you need, when you need it. Media is formulated without sodium bicarbonate to prevent clumping due to moisture absorption. Supplied in plastic bottles. Shipped at ambient temperature; recommended storage temperature: 2° to 8°C (36° to 46°F).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.