Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
22-363 Series
Fisherbrand™ Disposable Inoculating Loops and Needles
Ideal for use in laboratories
Designed for quantitative procedures such as sampling, serial dilutions, as well as for bacterial inoculation. Ideal for anaerobic microbiology applications where deep agar inoculation is required. Use in situations where flaming is not practical, such as under hoods and in anaerobic chambers. Flexible enough to facilitate streaking, yet rigid enough to penetrate the media for subsurface inoculation.
Includes: Certificate of dimensional accuracy
  • Precision molded from a specially formulated, biologically inert polymer
  • Combi Loop™ has a 10µL loop on one end and a 1µL loop on the other
  • More convenient than metal loops; use a loop directly from the package, then throw it away
  • Gamma irradiated for sterility in accordance with U.S. Pharmacopeia Volume XX
  • No flaming required, eliminating the hazards of open flames, spattering, cross-contamination, and the risk of infection from pathogens distributed by aerosol formation
  • Color coded for easy identification
  • Length: 9 in. (23cm)
View Specifications
Images for Fisherbrand™ Disposable Inoculating Loops and Needles
  • 22-363 Series
22-363 Series
22-363 Series
(6 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
22-363-602 Volume: 1µ to 10µL; Color: White
Pack of 500 for $79.89
Case of 10 PK for $688.45
Check Availability
Add
to Cart
(0) in My Cart
22-363-603 Volume: 1µL; Color: Blue
Case of 250 for $84.21
Check Availability
Add
to Cart
(0) in My Cart
22-363-604 Volume: 1µL; Color: Blue
Pack of 960 for $175.71
Case of 10 PK for $1,580.97
Check Availability
Add
to Cart
(0) in My Cart
22-363-605 Volume: 10µL, Color: Yellow; Individually wrapped; 250/PK.
Case of 250 for $87.96
Check Availability
Add
to Cart
(0) in My Cart
22-363-606 Volume: 10µL, Color: Yellow; 1000Pk
Case of 1000 for $184.14
Check Availability
Add
to Cart
(0) in My Cart
22-363-607 Volume: 10µL; Color: Yellow; 9600/Cs.
Pack of 960 for $177.09
Case of 10 PK for $1,591.33
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Description
  • Precision molded from a specially formulated, biologically inert polymer
  • Combi Loop™ has a 10µL loop on one end and a 1µL loop on the other
  • More convenient than metal loops; use a loop directly from the package, then throw it away
  • Gamma irradiated for sterility in accordance with U.S. Pharmacopeia Volume XX
  • No flaming required, eliminating the hazards of open flames, spattering, cross-contamination, and the risk of infection from pathogens distributed by aerosol formation
  • Color coded for easy identification
  • Length: 9 in. (23cm)