Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
View Image Gallery (6) Image(s) are representative of the product Group
  • TF7400, TF7401
  • TF7402, TF7403
  • TF7404, TF7405
  • TF7406, TF7407
Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ CryoPlus™ Storage Systems
Low-profile storage tanks combine liquid nitrogen storage reliability with microprocessor technology
Sixteen preset audible and visual alarm combinations provide maximum product protection. Automatic defogger is incorporated into the manual fill index to improve visibility during retrieval.
Includes: CryoPlus 1 has 0.5 in. 45° flare, 4 ft. (1.2m) flexible hose. CryoPlus 2, 3, and 4 come with 0.5 in. 45° flare, 6 ft. (1.8m) flexible hose.
Microprocessor Control
  • Precise liquid level control with over 16 parameters; with autofill system
  • Control panel is mounted on top for convenient access and easy touchpad programming
  • Digital temperature display sensor is located under the lid; this indicates the highest temperature reading inside the tank
  • 24 tricolor LEDs continuously display actual liquid nitrogen level and high-level/low-level setpoints
  • Alarm contacts for remote monitoring of storage tank conditions
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ CryoPlus™ Storage Systems
  • TF7400, TF7401
  • TF7402, TF7403
  • TF7404, TF7405
  • TF7406, TF7407
(6 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
TF7406
Thermo Scientific
No.:7406
552L, 100/120V
Required Accessories
Each for $24,887.50
Check Availability
Add
to Cart
(0) in My Cart
TF7400 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:7400
90L, 100/120V
Accessories
Each for $9,781.00
Check Availability
Add
to Cart
(0) in My Cart
TF7402 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:7402
200L, 100/120V
Required Accessories
Each for $13,257.80
Check Availability
Add
to Cart
(0) in My Cart
TF7404 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:7404
340L, 100/120V
Required Accessories
Each for $19,162.00
Check Availability
Add
to Cart
(0) in My Cart
TF7401
Thermo Scientific
No.:7401/PRC
90L, 200/230V
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
TF7405
Thermo Scientific
No.:7405/PRC
340L, 200/230V
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
Add Items to Cart
Description
Microprocessor Control
  • Precise liquid level control with over 16 parameters; with autofill system
  • Control panel is mounted on top for convenient access and easy touchpad programming
  • Digital temperature display sensor is located under the lid; this indicates the highest temperature reading inside the tank
  • 24 tricolor LEDs continuously display actual liquid nitrogen level and high-level/low-level setpoints
  • Alarm contacts for remote monitoring of storage tank conditions
Durable Construction
  • Low-profile tank with vacuum-insulated, stainless-steel chamber Counterbalanced lid with foamed-in-place, high density polyurethane insulation; dedicated vent; 100% clearance Two independent, flexible lid gaskets reduce moisture migration into the chamber
  • Liquid level sensor and fill port within vacuum space, for maximum storage and sensor protection
  • Front-mounted key lock and rear-mounted recessed power switch; control panel can also be locked
  • Temperature sleeve (included) provides colder temperatures and efficient vapor phase operation
  • Heavy-duty casters
Specifications & Ordering Information:
Optional racks, canisters, frames and risers allow you to create an organized inventory system that provides optimum storage in liquid or vapor phase. Contact your Customer Service Representative for details.
Compliance: UL listed to U.S. and Canadian requirements (100/120V units)
Specifications
Color White
Body18-gauge cold-rolled steel with powder-coated paint
ReservoirStainless steel
With Level MonitorYes
With Autofill SystemYes
† Add 5 in. (12.7cm) to length for utilities and lid opening. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.