Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
13-642-054
View Image Gallery (2) Image(s) are representative of the product Group
  • 13-642-054
  • Quick-Start Guide
Thermo Scientific™ Orion™ 3-Star Plus Conductivity Benchtop Meter
Dedicated measurement of conductivity, resistivity, salinity and TDS in a compact, ergonomic package
Includes: Meter Only 1114000 package includes meter, universal power supply and instructions.
Meter Kit 1114001 package includes meter, DuraProbe™ conductivity cell 013005MD, 5 x 60mL of 1413µS/cm conductivity/TDS standard 011007, electrode stand, universal power supply and instructions.
Meter Kit/Ultrapure Water 1114002 package includes meter, DuraProbe conductivity cell 013016MD, 5 x 60mL of 100µS/cm conductivity/TDS standard 011008, electrode stand, universal power supply and instructions.
Meter Kit/USP 645 1114003 package includes meter, DuraProbe conductivity cell 013016MD, 5 x 60mL of 100µS/cm conductivity/TDS standard 011008, electrode stand, Conductivity Calibration Resistor Kit, universal power supply and instructions.
Easy to operate in any mode
  • Bright, backlit LCD display with ready indication
  • Simultaneous display of conductivity and temperature
  • Convenient built-in Quick-Start Guide with calibration instructions
  • Stores, displays and prints up to 10 methods with individual method password protection
  • Automatic or manual temperature compensation
  • Auto-read/hold function
  • Capable of operating an autosampler and stirrer probe (both sold separately)
  • IP54 dustproof and splashproof
Extra features for Conductivity measurements
  • 1 to 5 calibration points
  • Three decimal point cell constant entry
  • Compatible with 2-cell, 4-cell, planar cell probes
  • Reference temperature: 5°C, 10°C,15°C, 20°C or 25°C
  • Linear or nonlinear temperature compensation
View Specifications
Images for Thermo Scientific™ Orion™ 3-Star Plus Conductivity Benchtop Meter
  • 13-642-054
  • Quick-Start Guide
13-642-054
13-642-054
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
13645104
THERMO SCI ORION
No.:1114001-WA
Meter kit with 5 x 60mL 1413µ S/cm standard
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Easy to operate in any mode
  • Bright, backlit LCD display with ready indication
  • Simultaneous display of conductivity and temperature
  • Convenient built-in Quick-Start Guide with calibration instructions
  • Stores, displays and prints up to 10 methods with individual method password protection
  • Automatic or manual temperature compensation
  • Auto-read/hold function
  • Capable of operating an autosampler and stirrer probe (both sold separately)
  • IP54 dustproof and splashproof
Extra features for Conductivity measurements
  • 1 to 5 calibration points
  • Three decimal point cell constant entry
  • Compatible with 2-cell, 4-cell, planar cell probes
  • Reference temperature: 5°C, 10°C,15°C, 20°C or 25°C
  • Linear or nonlinear temperature compensation
Increased accuracy
  • Improved stability algorithm improves calibration and measurement accuracy
  • SMART STABILITY™ and SMART AVERAGING™ functions optimize accuracy, precision and response time
  • ATC calibration
  • Advanced self-tests on meter operation, self-diagnostics
Complete documentation
  • Full complement of GLP-related features
  • 1000 point datalogger, downloadable to printer or computer via RS-232 connection
  • Average slope and slope of segment printed with measurement
Certifications: CE, CSA, UL TUV and FCC Class A limits approved
Compliance: 21 CFR Part II Compliant with optional software. Meets all U.S. Pharmacopeia (USP) requirements.
Recommended Storage:
Warranty: Three years
Specifications
ModelOrion 3-Star Plus Conductivity Benchtop Meter
Conductivity Mode
Range0.000 to 3000mS/cm, auto resolution with cell constant dependence
Resolution4 significant digits down to 0.001µS/cm, dependent on cell constant
Relative Accuracy±0.5% ±1 digit or 0.01µS/cm, whichever is greater
Cell Constants0.001 to 199.9cm-1
Cell Compatibility2-Cell, 4-Cell, or Planar
CalibrationDirectCal™/AUTO-CAL™: 1 to 5 points/1 to 3 points
Reference Temperature5°C, 10°C,15°C, 20°C or 25°C (default); autoranging
Resistivity Mode
Range0.0001 to 100megohm
ResolutionAutoranging
Relative Accuracy±0.5% ±1 digit
Salinity Mode
Range0.1 to 80.0ppt NaCl equivalent; 0.1 to 42ppt practical
Resolution0.1ppt
Relative Accuracy±0.1ppt ±1 digit
TDS Mode
Range0 to 19,999mg/L
Resolution1mg/L
Relative Accuracy±0.5% ±1 digit
Adjustable TDS Factor0.0 to 10.0%
Temperature Mode
Range-5° to +105°C
Resolution0.1°C up to 99.9°C, 1.0 over 99.9°C
Relative Accuracy±0.1°, ±1 digit
Temperature CompensationnLF, linear
DisplayCustom LCD; simultaneous display of selected measurement and temperature, and current electrode condition
Inputs/Outputs
Inputs8-Pin Mini-DIN Locking for Conductivity; Phono jack for stirrer
Sensor Types2-Cell, 4-Cell, or Planar Conductivity
OutputsBidirectional RS-232, stereo jack, optional USB inteface cable
Data ManagementGLP Electrode Veriļ¬cationSaves 1000 points by time, stability or manual read with time/date stampCalibration log of last 10 calibrationsUser method storage for 10 proceduresAdvanced self-test and diagnostics routinesPassword protectionDownloadable software and methods updates via Internet
Regulatory and Safety Compliance 
Environmental Operating Conditions
Ambient Operating Temperature5° to 45°C
Relative Humidity5 to 85% noncondensing
IP RatingIP54 compliance: splashproof and dustproof
StabilitySMART STABILITY™ and SMART AVERAGING™: Resolution dependent/automatic
Power SupplyUniversal AC power adapter or 4 x 1.5V AA (optional)
WarrantyThree years from date of purchase