Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
13-642-059
View Image Gallery (2) Image(s) are representative of the product Group
  • 13-642-059
  • Waterproof Connector
Thermo Scientific* Orion* 3-Star Plus pH Portable Meter
Waterproof, battery-operated meter stands up to the most demanding field applications
Stands up to the most demanding quality control and general analysis applications and delivers over 2000 hours of continuous operation. Improved stability algorithm to increase calibration and measurement accuracy.
Includes: Meter Only 1212000 package includes meter; batteries and instructions.
Meter Kit/pH 1212001 package includes meter; pH/ATC Triode* 9107BNMD; 10 x 15mL pouches each of pH 4.01/7.00/10.01 buffers; 5 x 60mL pH electrode storage solution; beaker; electrode storage chamber; hard field case; batteries and instructions.
Meter Kit/pH 1212002 package includes meter; pH/ATC NoCal* electrode 5107BNMD; 10 x 15mL pouches each of pH 4.01/7.00/10.01 buffers; 5 x 60mL pH electrode storage solution; beaker; hard field case; batteries and instructions.
Meter Kit/pH 1212005 package includes meter; pH/ATC Triode* 8107BNMD; 10 x 15mL pouches each of pH 4.01/7.00/10.01 buffers; 5 x 60mL pH electrode storage solution; beaker, electrode storage chamber, soft field case; batteries and instructions.
Meter Kit/pH 1212503 package includes meter; ORP/ATC Triode* 9179BNMD; 5 x 60mL ORP Standard; meter impact jacket with electrode storage chamber, soft field case; batteries and instructions.
Meter Kit/pH 1212101 package includes meter; pH/ATC Triode* 9107BNMD; batteries and instructions.
Easy to operate in any mode
  • Bright, backlit LCD display with ready indication
  • Balanced comfortable handling
  • Simultaneous display of pH or mV and temperature
  • Convenient built-in Quick-Start Guide with calibration instructions
  • Stores, displays and prints up to 10 methods with individual method password protection
  • Automatic or manual temperature compensation
  • Auto-read/hold function
  • IP67 waterproof
Extra features in every mode
  • pH: automatic or manual calibration; 1 to 5 calibration points; auto-buffer-recognition of NIST*/US and DIN buffers
  • mV/ORP: EH autocalibration to standard hydrogen electrode
View Specifications
Alert: Products on this page may include voltages not considered standard in North America. Please review carefully.
Images for Thermo Scientific* Orion* 3-Star Plus pH Portable Meter
  • 13-642-059
  • Waterproof Connector
13-642-059
(7 item(s) in product Group)
  • Items
  • Specifications
Description Catalog No. Availability Quantity Price
Kit w/pH/ATC triode; 3-Star Plus
13650139
THERMO SCI ORION
No.:1212101
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
For 3-Star portable pH meter
13645112
THERMO SCI ORION
No.:1212002
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
For 3-Star pH portable meter
13645111
THERMO SCI ORION
No.:1212001
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Kit w/pH/ATC triode; 3-Star Plus; w/10 x 15mL buffer
13650140
THERMO SCI ORION
No.:1212501
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Kit w/ ORP/ATC triode; 3-Star Plus; 5 x 60mL ORP standard
13650141
THERMO SCI ORION
No.:1212503
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Orion 3-Star Plus pH Portable Meter Only 1212000; includes: meter; batteries and instructions
13-642-059
Thermo Scientific
No.:1212000
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
pH buffer pouches (4, 7 and 10) and hard field case
Accessories
13-302-223
Thermo Scientific
No.:1212005
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Add Items to Cart
Description
Easy to operate in any mode
  • Bright, backlit LCD display with ready indication
  • Balanced comfortable handling
  • Simultaneous display of pH or mV and temperature
  • Convenient built-in Quick-Start Guide with calibration instructions
  • Stores, displays and prints up to 10 methods with individual method password protection
  • Automatic or manual temperature compensation
  • Auto-read/hold function
  • IP67 waterproof
Extra features in every mode
  • pH: automatic or manual calibration; 1 to 5 calibration points; auto-buffer-recognition of NIST*/US and DIN buffers
  • mV/ORP: EH autocalibration to standard hydrogen electrode
Increased accuracy
  • Improved stability algorithm improves calibration and measurement accuracy
  • SMART STABILITY* and SMART AVERAGING* functions optimize accuracy, precision and response time
  • ATC calibration
  • Advanced self-tests on meter operation, self-diagnostics
Complete documentation
  • Full complement of GLP-related features
  • 1000 point datalogger, downloadable to printer or computer via RS-232 connection
  • Average slope and slope of segment printed with measurement
Certifications: CE, CSA, UL and FCC Class A limits approved
Compliance: 21 CFR Part II Compliance with optional software
Warranty: Three years
Specifications
ModelOrion 3-Star Plus pH Portable Meter
pH Mode
Range-2.000 to +19.999pH
Resolution0.1/0.01/0.001pH
Relative Accuracy±0.002pH
CalibrationAUTO-CAL* and/or Manual Calibration; 1 to 5 points; US/NIST, DIN, and custom buffers recognized
mV Mode
Range±1999.9mV
Resolution0.1mV
Relative Accuracy±0.2 mV or 0.05%, whichever is greater
ORP CalibrationEH autocalibration to standard hydrogen electrode
Temperature Mode
Range-5° to +105°C
Resolution0.1°C up to 99.9°C, 1.0°C over 99.9°C
Relative Accuracy±0.1°, ± 1 digit
CompensationAutomatic or manual, -5° to +105°C
DisplayCustom backlit LCD; simultaneous display of pH or mV and temperature, pH calibration points, and current electrode condition
Inputs/Outputs
InputsWaterproof BNC for pH/ORP electrodes; waterproof 8-Pin Mini-DIN Locking for ATC probe; Pin tip for separate reference electrode
Sensor TypepH/ORP Electrode
OutputsWaterproof bidirectional RS-232, stereo jack, optional USB interface cable
Data ManagementGLP Electrode VerificationSaves 1000 points by time, stability or manual read with time/date stampCalibration log of last 10 calibrationsUser method storage for 10 proceduresAdvanced self-test and diagnostics routinesPassword protectionDownloadable software and methods updates via Internet
Regulatory and Safety Compliance 
Environmental Operating Conditions
Ambient Operating Temperature5° to 45°C
Relative Humidity5 to 85% noncondensing
IP RatingIP67 waterproof
StabilitySMART STABILITY* and SMART AVERAGING*: Resolution dependent/automatic
Power Supply4 x 1.5V "AA" batteries
Warranty PeriodThree years from date of purchase