Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
View Image Gallery (6) Image(s) are representative of the product Group
GE* Healthcare HiTrap* IEX FF Columns
For method scouting, group separations, concentrations and sample cleanup
  • Convenient for rapid separations either alone or connected in series
  • Designed for use with syringe, peristaltic pump or chromatographic system
  • High flowrates and good scale-up potential
View Specifications
Images for GE* Healthcare HiTrap* IEX FF Columns
(10 item(s) in product Group)
  • Items
  • Specifications
Description Catalog No. Availability Quantity Price
Prepacked w/Q Sepharose FF; 5 x 1mL
450-002-58
GE Healthcare
No.:17-5053-01
N/AYou must be logged in and have a valid account number to view item availability   Pack of 5 for $140.68
(0) in My Cart
Prepacked w/SP Sepharose FF, strong cation; 5 x 1mL
450-002-59
GE Healthcare
No.:17-5054-01
N/AYou must be logged in and have a valid account number to view item availability   Pack of 5 for $140.68
(0) in My Cart
Prepacked w/DEAE Sepharose FF, weak anion; 5 x 1mL
450-002-60
GE Healthcare
No.:17-5055-01
N/AYou must be logged in and have a valid account number to view item availability   Pack of 5 for $140.68
(0) in My Cart
Prepacked w/CM Sepharose FF, weak cation; 5 x 1mL
450-002-61
GE Healthcare
No.:17-5056-01
N/AYou must be logged in and have a valid account number to view item availability   Pack of 5 for $140.68
(0) in My Cart
Prepacked w/DEAE Sepharose FF, weak anion; 5 x 5mL
450-002-91
GE Healthcare
No.:17-5154-01
N/AYou must be logged in and have a valid account number to view item availability   Pack of 5 for $304.03
(0) in My Cart
Prepacked w/CM Sepharose FF, weak cation; 5 x 5mL
450-002-92
GE Healthcare
No.:17-5155-01
N/AYou must be logged in and have a valid account number to view item availability   Pack of 5 for $304.03
(0) in My Cart
Prepacked w/Q Sepharose FF; 5 x 5mL
450-002-93
GE Healthcare
No.:17-5156-01
N/AYou must be logged in and have a valid account number to view item availability   Pack of 5 for $304.03
(0) in My Cart
Prepacked w/SP Sepharose FF, strong cation; 5 x 5mL
450-002-94
GE Healthcare
No.:17-5157-01
N/AYou must be logged in and have a valid account number to view item availability   Pack of 5 for $304.03
(0) in My Cart
Prepacked w/ANX Sepharose 4 Fast Flow (high sub); 5 x 1mL
450-002-99
GE Healthcare
No.:17-5162-01
N/AYou must be logged in and have a valid account number to view item availability   Pack of 5 for $140.68
(0) in My Cart
Prepacked w/ANX Sepharose 4 Fast Flow (high sub); 5 x 5mL
450-003-00
GE Healthcare
No.:17-5163-01
N/AYou must be logged in and have a valid account number to view item availability   Pack of 5 for $304.03
(0) in My Cart
Add Items to Cart
Description
  • Convenient for rapid separations either alone or connected in series
  • Designed for use with syringe, peristaltic pump or chromatographic system
  • High flowrates and good scale-up potential