Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
567-0020
Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with PES Membrane
1000mL; Pore Size: 0.20µm; Membrane: 90mm
Catalog No.: 09-741-03 Available on GSA/VA Contract for Federal Government customers only. 
09-741-03 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:567-0020
Unit, Filter; Thermo Scientific Nalgene; Rapid-Flow; all-in-one; polystyrene housing; cellulosic-plugged side arm; tubing adapter; graduated; PES; Capacity: 1000mL upper, 1000mL receiver; pore size: 0.2µm; membrane dia.: 90mm
Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number? Learn More
Price:
Case of 12 for $269.85
  Check Availability
Add
to Cart
(0) in My Cart
Includes: Polystyrene cover; graduated upper chamber and storage bottle (except 115 mL size); integral membrane filter; cellulosic-plugged side arm; quick-disconnect tubing adapter.
Images for Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with PES Membrane
  • 567-0020
567-0020
567-0020