Log into Your Account
Don't have a profile? Register
Forgot Password?
Search
Customer Service:  800-766-7000Web Support:  877-885-2081All Other Contact Info
Browse Results: 367 item(s)
You're Browsing:N - Z
  1. 1
  2. 2
  3. 3
  4. 4
  5. 5
  6. >
  7. >>
1 Fisher HealthCare* Sure-Vue* RPR Test Kit
Variation of the standard VDRL   ·  CLIA Complexity: Non-Waived   ·  CPT Codes: 86542/86593   ·  Cardiolipin charcoal microparticle test for the qualitative and semi-quantitative determination of reagin, which
2 Fisher HealthCare* Sure-Vue* SELECT Rubella Latex Test
Does not require a dilution step for 10 IU sensitivity   ·  CLIA Complexity: Non-Waived  ·  CPT Codes: 86762/86406  ·  Sensitivity: 100%  ·  Specificity: 98.6%  ·  Rapid latex agglutination
3 Fisher HealthCare* Sure-Vue* RSV Test Kit Controls
For use with Sure-Vue RSV Test Kit Respiratory Syncytial Virus (RSV)
4 Siemens* Multistix PRO* Reagent Strips
Two proprietary dip-and-read tests, Protein Low (Albumin) and Creatinine   ·  CLIA Complexity: Waived   ·  Results in 60 seconds Provide protein-to-creatinine ratio Compatible with Clinitek* analyzers  ·  Improved protein
5 Fisherbrand* Urine Reagent Strips
Read visually or use with the AimStrip* Urine Analyzer and CTMI CT-120 Urine Analyzer   ·  Functionally equivalent to Siemens* Multistix* 10-SG's  ·  Ten parameters that are in the same sequence as Siemens Multistix 10-SG's  ·  Reliable
6 Fisherbrand* Urine Sediment Tubes
Designed for use with Sysmex* and Siemens* Atlas* instruments   ·  Non-flared top for efficient nesting in tube racking systems  ·  Circular in shape  ·  Constructed of shatter-resistant polystyrene  ·  Transparent
7 Fisher HealthCare* Sure-Vue* Rubella Latex Test Kit
Requires a dilution step for 10 IU sensitivity   ·  CLIA Complexity: Non-Waived  ·  CPT Codes: 86762/86406  ·  Sensitivity: 99.5%   ·  Specificity: 96.1%  ·  Non-dilution: 1-2 IU sensitivity;
8 Fisher HealthCare* Sure-Vue* Rota Test Kit
Simple two-step procedure   ·  CLIA Complexity: Non-Waived  ·  CPT Code: 87425  ·  Rapid membrane-based immunogold assay for the qualitative detection of rotavirus antigen in human fecal specimens; aids in the
9 Fisher HealthCare* Sure-Vue* Strep A Lateral Flow Test Kit
One-step lateral-flow immunoassay for the qualitative detection of Group A Strep from throat swabs   ·  CLIA Complexity: Waived   ·  CPT Code: 87081  ·  Sensitivity: 97%  ·  Specificity: 95%  ·  Intended
10 Fisher HealthCare* Sure-Vue* RSV Test Kit
Alternative procedure for bloody and mucoid samples   ·  CLIA Complexity: Waived and Moderate  ·  CPT Codes: 87807QW, 87807  ·  Sensitivity: >99%  ·  Specificity: >99%  ·  Results
11 Fisher Diagnostics Pacific Hemostasis* SickleScreen* Assay Kit
Determines the concentration of Hemoglobin S in whole blood samples The Thermo Scientific SickleScreen products use a modified Nalbandian procedure that provides fast and accurate results for screening hereditary Sickle Cell disease and Sickle Cell Trait. Both Sickle Cell disease and Sickle Cell trait
12 SA Scientific* StrepAlert* (Dipstick) Strep A Test Kit
Membrane immunoassay kit with results in as little as five minutes   ·  CLIA Complexity: Waived and Non-waived  ·  CPT Codes: 87880QW (Waived); 87880 (Non-waived)  ·  Sensitivity: 97.9%  ·  Specificity:
13 Fisher HealthCare* Sure-Vue* SELECT Staph Latex Test Kit
  ·  CLIA Complexity: Non-Waived  ·  CPT Code: 87077  ·  Sensitivity: 99%  ·  Specificity: 92%  ·  Latex agglutination slide test for the qualitative detection of Staphylococcus
14 Fisherbrand* Urine Sedimentation System
Designed to produce the same results as the most popular style urine sedimentation system using the same test procedure   ·  15mL graduated urine tube in economical or break-resistant polystyrene  ·  Decanting pipet allows supernatant to be decanted off; leaves 1mL
15 Fisher HealthCare* Sure-Vue* RF Rapid Latex Test Kit
Suspension of latex particles is coated with human immunoglobulin   ·  CLIA Complexity: Non-Waived  ·  CPT Codes: 86430/86318  ·  Latex agglutination test for the qualitative and semiquantitative determination
  1. 1
  2. 2
  3. 3
  4. 4
  5. 5
  6. >
  7. >>