Log into Your Account
Don't have a profile? Register
Forgot Password?
Search
Customer Service:  800-766-7000Web Support:  877-885-2081All Other Contact Info
Browse Results: 386 item(s)
You're Browsing:Syringes and Needles
  1. 1
  2. 2
  3. 3
  4. 4
  5. 5
  6. >
  7. >>
1 Surgical Design Innoculating needles
Innoculating needle
2 Hamilton Company 7000 Series Modified MICROLITER* Syringes
The 7000 series is the only zero dead volume syringe, ideal for dispensing volumes from 0.05µL up to 5µL Considered modified MICROLITER syringes due to their unique design. The fine gauge plunger wire travels to the tip of the needle ensuring that all solvent is expelled from the needle. 7000
3 Hamilton Company 1800 Series GASTIGHT Syringes
Reinforced plunger version of the 1700 series syringes is ideal for dispensing volumes from 1 μL up to 500 μL. Attached to the flange of the syringe is a blue Syringe Holder. The Syringe Holder provides a spot to hold the syringe that will not heat up the barrel causing dispense inaccuracies. Additionally
4 Restek Presents Hamilton Standard Microliter Volume Syringes
5 SGE* Replacement Plungers
For SGE replaceable-plunger syringes listed. For additional replacements, contact your Fisher Scientific Customer Service Representative. For syringe models SG-003987, SG-003988; 25 microliter capacity 1mL capacity For syringe models SG-008900, SG-008960, SG-008970, SG-008975; 10mL capacity Plng Rpl
6 Hamilton Company 1700 and 1000 Series GASTIGHT* SampleLock* Syringes
Hamilton SampleLock syringes allow you to collect, store, transport and analyze liquid or gaseous samples without the danger of evaporative loss or environmental contamination. Syringes are available in sizes ranging from 50µL to 100mL. An easy-to-use twist valve and a positive rear stop on SampleLock
7 Smiths Medical Saf-T Holder* Blood Culture Holders and Transfer Devices
Come with back-end needle and multi-sample Luer permanently attached to the holder   ·  Eliminate potential exposures and holder reuse  ·  Provide safe needleless transfer after a syringe draw  ·   With female luer With male luer
8 Thermo Scientific* National* Target* Precision LC Syringes for Waters* Injection Valves
Use with U6K manual injector   ·  LC manual injection valve syringes  ·  Removable needle syringes permit easy removal of bent needles for replacement; same syringe can be used with different gauge needles to meet specific application needs  ·  PTFE-tipped
9 Hamilton Company 700 Series MICROLITER* Syringes
Original hand-fitted Hamilton syringe. Ideal for dispensing volumes from 0.5 up to 500µL. The stainless-steel plunger is manufactured to fit the glass barrel with a tolerance smaller than 100 millionths of an inch, resulting in unsurpassed syringe life. 700 Series Syringe Terminations
10 Terumo Hypodermic Syringes
W/O NDL 21G X 1 in. W/O NDL 3mL W/O NDL 22GX1IN W/O NDL 25GX5/8IN W/O NDL W/O NDL W/O NDL
11 BD Vacutainer* Slip Tip Syringe
10 cc, slip tip
12 Hamilton Company 700, 1700 and 7000 Series GASTIGHT* and MICROLITER* Digital Syringes
Easily achieves precision up to 0.5% while dispensing volumes from 500µL down to 50nL Syringe eliminates parallax errors by providing a digital readout on its integrated LCD screen. An adjustable plunger-stop guarantees reproducible plunger placement. Easy-To-Read Read the syringe
13 Thermo Scientific SUN-SRi* Luer-Slip Syringes
Offered in a range of sizes, from 1mL to 50mL, sterile and non-sterile   ·  Chemically resistant, inert polypropylene and polyethylene  ·  No rubber plunger seals or silicone lubricants that may cause sample contamination  ·  Bulk packaged, non sterile-no
14 Hamilton Company 1700 Series Gastight* Syringes
Gastight version of the original 700 series syringes, ideal for dispensing volumes from 1 up to 500µL Syringe series offers a variety of different terminations and needle options. 1700 Series Syringe Terminations   ·  Cemented Needle (N)- For low volume
15 Thermo Scientific* Gilmont* Micrometer Syringes and Burets
High precision with economy of cost   ·  0.5% accuracy  ·  Liquid comes in contact with only glass, VITON* and PTFE*  ·  VITON O-ring gives a vacuum tight seal  ·  All other plastic parts of polypropylene—instrument
  1. 1
  2. 2
  3. 3
  4. 4
  5. 5
  6. >
  7. >>