Log into Your Account
Don't have a profile? Register
Forgot Password?
Search
Customer Service:  800-766-7000Web Support:  877-885-2081All Other Contact Info
Browse Results: 262 item(s)
You're Browsing:Blood Collection Supplies
  1. 1
  2. 2
  3. 3
  4. 4
  5. 5
  6. >
  7. >>
1 Green Product Fisherbrand* Microhematocrit Capillary Tubes
Low-taper wall construction guarantees error less than 1% absolute, 0.4 hematocrit, relative   ·  Ends can be sealed by sealing clay or flame  ·  Manufactured from low-taper, soft glass  ·  Red band on tube indicates heparinization; blue band indicates
2 Propper Select* Micro-Hematocrit Capillary Tubes
For collection and transfer of blood. Glass tubes are alkali-free and non-corrosive. 75mm x 1.4mm
3 Fisherbrand* IB* Serum Filter
Allows for direct instrument sampling   ·  For use with 13mm plastic evacuated blood collection tubes  ·  Store plasma up to four days, serum up to seven days when it is essential to prevent any exchange of ions  ·  One-way valve permits leak resistant
4 Fisherbrand* Blood Bands with Self-Adhesive Tails
Red
5 Fisherbrand SQ-EASY* Prefilter System Components
Disposable and efficient self-contained filtration system   ·  For use in stool, serum, diluted whole blood, urine, and throat swab assays  ·  Components are liquid-tight and interlocking to prevent sample leakage  ·  Range of filtration materials available,
6 Fisherbrand* Plastic Microhematocrit Tubes
Clarity and performance of glass—shatterproof safety of plastic   ·  No need to perform correlation studies  ·  Available untreated and heparinized  ·  Eliminates the risk of transmission of bloodborne pathogens associated with broken glass tubes  ·  Better
7 Fisherbrand* Safety Lancets
Built-in needle control depth (1.8mm) for consistent and accurate sampling results   ·  Sterile lancet needle is hidden inside the cartridge and automatically retracts after use  ·  Helps protect patient and health professional against accidental needlestick injuries
8 Fisherbrand* Standard Serum Filters
Used when the filtered serum can be immediately poured off for analysis Disposable devices for filtering fibrin from the serum portion of centrifuged blood samples contained in standard 16-, 13- or 10.25-mm I.D. collection tubes   ·  Standard model is
9 Fisherbrand* Unistik* 2 Disposable Fingerstick Lancet Devices
Ideal for eliminating accidental needlesticks during capillary sampling   ·  Deliver optimum blood flow quickly without sacrificing patient comfort  ·  Disposable capillary sampling devices designed for one-time use  ·  Simple to use: push the lancet into
10 Fisherbrand* IB* Model Serum Filters
Store serum up to seven days when it is essential to prevent any exchange of ions   ·  One-way valve permits a physical gap to be created between the serum/plasma and clot, and prevents backflow into the primary collection tube  ·  Use with glass or plastic collection
11 Fisherbrand* DISPENSE Filter System
Saves time, reduces lab costs and increases safety The restricted opening provides effective specimen closure and can help minimize exposure to droplets of blood or other potentially infectious materials during sample handling, preparation, processing, and storage.   ·  All-inclusive
12 Green Product Fisherbrand* Color-Coded Capillary Tubes
Excellent for use in microhematocrit determinations   ·  Ends can be sealed by flame, sealing clay or caps  ·  Manufactured from low-taper, soft glass  ·  Tubes are packed in plastic vials with reclosable snap-lock caps to maintain cleanliness  ·  Red
13 Fisherbrand* Infant Heel Warmers
The easiest, safest, and most reliable way to obtain better heelsticks The increased blood flow makes fewer skin punctures necessary, which allows for less squeezing, fewer infections, less risk of cartilage damage and faster healing.   ·  Provide safe, nontoxic heat in a format
14 Green Product Fisherbrand* Micro Blood Collecting Tubes
With tapered bottom tips to facilitate micro blood sample collection   ·  For Natelson and Caraway techniques  ·  Color-coded tops distinguish between plain and heparinized tubes  ·  Heparinized tubes are treated with ammonium heparin  ·  Natelson
15 Fisherbrand* Natural Latex Disposable Tourniquets
Smooth surface for easy tying and untying   ·  Made of the highest-quality latex  ·  Disposable  ·  Measures 18 x 1 in. (45.7 x 2.5cm)  ·  Thickness 0.03 in. (0.07cm)  ·   Natural latex; disposable
  1. 1
  2. 2
  3. 3
  4. 4
  5. 5
  6. >
  7. >>