Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Browse Results: 268 item(s)
You're Browsing:Stirrers
  1. 1
  2. 2
  3. 3
  4. 4
  5. 5
  6. >
  7. >>
1 Available on GSA/VA Contract for Federal Government customers only. Fisherbrand* Magnetic Stir Bars with Vanes
Ensure thorough sample mixing in test tubes, micro vials and conical-bottom centrifuge tubes   ·  PTFE that encases magnetic bars also forms solid, flat stirring vane  ·  Permit rotation in small conical tubes and can be trimmed to fit without affecting the magnet  ·  
2 Available on GSA/VA Contract for Federal Government customers only. Fisherbrand* Polygon Stir Bars
Available in five sizes   ·  Smooth polygon bars  ·  The FDA-grade PTFE of these stirring bars is low-friction, inert and durable  ·  Alnico magnet (Ni, Ni, Fe and Co)  ·  Suitable for general stirring applications  ·  Manufactured
3 Available on GSA/VA Contract for Federal Government customers only. Fisherbrand* Magnetic Stir Bar Six Packs
Our most popular sizes in convenient, economical, ready-to-use multipacks   ·  FDA-grade PTFE coating  ·  Smooth, seamless, polished finish  ·  High-strength magnetization  ·  Micro bars or octagonal bars  ·  Octagonal bars have
4 Available on GSA/VA Contract for Federal Government customers only. Fisherbrand* Round Stir Bars with Removable Pivot Ring
PTFE round bars offer excellent centering and smooth running characteristics   ·  Small removable pivot ring in the center adds to their versatility  ·  Pivot ring minimizes contact area of bar to vessel  ·  Ring also reduces friction and the marring of
5 Available on GSA/VA Contract for Federal Government customers only. Fisherbrand* Octagonal Magnetic Stir Bars
Flat surfaces provide greater mixing action   ·  With pivot rings for reduced friction and chattering  ·  Alnico-V (Al, Ni, Fe and Co) magnets encased in PTFE fluoropolymer  ·  Octagonal shape offers more surface area and greater turbulence than does a
6 Fisherbrand Micro Stir Bars
  ·  Octagonal shape  ·  Resists corrosion, heat, and cold  ·  Available in blue, red or yellow  ·  PTFE  ·   Micro Yellow, 10 x 2mm Micro Blue, 7 x 2mm Micro Yellow, 8 x 1.5mm Micro Red, 7 x 2mm Micro Yellow, 10 x 2mm Micro
7 Fisher Scientific* Twistir* Mini Stirrers
Stir with style – economical personal stirrers with a unique triangular design Choice of seven whimsical top plate designs.   ·  Stirring range: 350 to 2000rpm  ·  Stirring capacity: 1L  ·  Maximum vessel diameter: 8.5cm  ·  Power-on
8 Fisherbrand* Stirrer Anchors
Anchor type blade Inert, unbreakable, and will not scratch Useable to 260°C Shaft Diameter: 6mm 500mm 400mm
9 Fisherbrand* Stirring Paddles
Ideal for use in stirring small volumes or for chemical mixing in photographic work Choose from small or large size paddles.   ·  Made from high-density polyethylene  ·  Small paddle is ideal for use in stirring small volumes or for chemical mixing in photographic work  ·  Large
10 Available on GSA/VA Contract for Federal Government customers only. Fisherbrand* 8-Agon Stir Bars
Octagon-shaped, PTFE stir bars with molded-on pivot ring   ·  Interrupted surface provides greater surface area and added turbulence  ·  Molded pivot ring aids in reducing friction and chattering  ·  Autoclavable  ·   Red; 8 dia. x 22mm
11 Available on GSA/VA Contract for Federal Government customers only. Fisherbrand* Egg-Shaped Bars
Designed for use in round-bottom vessels   ·  Pivot point integral to the design without a separate ring or abrupt change in contour  ·  Covered with PTFE  ·  Shape mimics that of the flask for complete mixing  ·  Spin unrestricted by vessel
12 Available on GSA/VA Contract for Federal Government customers only. Fisherbrand Soft Glass Stirring Rods
Available in different lengths ranging from 5 to 10 in.   ·  With rounded and fire-polished ends  ·   Diameter: 0.18 in. (4.5mm); Length: 8 in. (2.3mm) Dia. x L: 0.19 in. x 6 in. Dia. x L: 0.125 in. x 5 in. Stirring rod; Diameter: 0.25 in. (6.3mm); Length: 10
13 Products for Science Education Mini-Stirrer
Large diameter top minimizes spill risks, making it ideal for student use.   ·  Variable speed from 0 to 1650rpm   ·  Front-mounted, stepless control knob   ·  Strong aluminum top plate with baked enamel finish   ·  Large 12cm diameter stirring surface reduces spill risks   ·  Includes
14 Products for Science Education Fisher Scientific* Ceramic-Top Magnetic Stirrers
Sleek, low-profile stirrers.   ·  Variable speed control from 100 to 1400rpm   ·  Instant-on feature ensures uniform low-speed stirring at 100rpm  ·  Quiet-running, direct-drive motor  ·  Reflective, porcelain-coated stainless-steel top surface
15 Available on GSA/VA Contract for Federal Government customers only. Fisherbrand* Magnetic Micro Stirring Bars
For stirring in test tubes, micro beakers   ·  Particularly useful in disciplines including life sciences and environmental testing, in which small sample volumes need to be prepared and evaluated  ·  Covered with PTFE fluoropolymer  ·  Each bar packaged
  1. 1
  2. 2
  3. 3
  4. 4
  5. 5
  6. >
  7. >>