Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Browse Results: 15 item(s)
You're Browsing:Centrifugal Filter Devices
from Viva Products + Iris Sample Processing + Thermo Scientific Anatomical Pathology + Fisher Scientific + Thermo Scientiific Sun SRI
  1. 1
1 Available on GSA/VA Contract for Federal Government customers only. Fisherbrand* Filter Cards for Shandon* Cytospin*
White cards are made from a blend of 100% cotton and cellulose fibers   ·  Excellent absorbent padding for the collection of excess reagents or wash solution  ·  Cellulose fibers improve the card's strength and durability during centrifugation and facilitate the
2 Available on GSA/VA Contract for Federal Government customers only. Fisherbrand* Single Cytology Funnels
For the Shandon* CytoSpin* Centrifuge   ·  Functional equivalent of the Shandon Single Cytofunnel*  ·  Fully compatible with Shandon CytoSpin Centrifuge  ·  White Filter Card is pre-attached and is designed for single use only  ·  Brown
3 Fisherbrand* Disposable Filter Concentrators
Ensure consistent high-quality slide preps Save time and improve results by eliminating the need to clean and sterilize reusable sample chambers   ·  Cells from body fluids are concentrated onto microscope slides with a level of cell recovery, clarity, and detail that cannot be
4 Available on GSA/VA Contract for Federal Government customers only. Fisherbrand* Double Cytology Funnel with White Filter Cards
For the Shandon* CytoSpin* Centrifuge   ·  Functional equivalent of the Shandon Double Cytofunnel*  ·  Fully compatible with Shandon CytoSpin Centrifuge  ·  White filter card is pre-attached and is designed for single use only  ·  Funnel
5 Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific* Shandon* Single Cytofunnel* with Brown Filter Cards
Used for slower absorption of scanty hypocellular specimens   ·  Disposable sample chamber with brown pre-attached filter card and cap  ·  For single use with all Thermo Scientific Shandon Cytospin* Cytocentrifuge applications  ·  Eliminates the
6 Thermo Scientific* SUN-SRi* Centrifugal Filter Devices
Combination filter/receiver tube unit for sample processing directly in centifuges   ·  Units available for sample sizes from 200µL to 25mL  ·  For use in microcentrifuges, and benchtop and floor model centrifuges  ·  Choose from a variety of membrane
7 Thermo Scientific* Shandon* Double Cytofunnel*
For single use with all Thermo Scientific Shandon Cytospin* Cytocentrifuge applications and immunochemistry techniques Produce more cost-effective, thin-layer preparations when compared to other thin-layer methods   ·  Optimize cell yield   ·  Reduces the risk
8 Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific* Shandon* EZ Single Cytofunnel* with White Filter Cards and Caps
Disposable sample chamber with white filter card and cap   ·  For single use with all Thermo Scientific Shandon Cytospin* Cytocentrifuge applications  ·  Optimize cell yield  ·  Reduces the risk of exposure to pathogenic samples  ·  Disposable
9 Thermo Scientific* Shandon* Cytoblock* Cell Block Preparation System
Increases ability to process scanty or unseen material for cell block and reduces loss of critical material   ·  Increases ability to process scanty or unseen material for cell block and reduces loss of critical material  ·  Suitable for immunohistochemical techniques  ·  Replaces
10 Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific* Shandon* EZ Megafunnel* with Caps and Slides
  ·  Cytofunnels designed for spinning large samples up to 6mL  ·  Minimizes the need to pre-centrifuge  ·  Cell deposition area of 22 x 14.75mm (325mm2) for increased cell yield  ·  Prepares both air-dried and fixed preparations,
11 Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific* Shandon* EZ Single Cytofunnel* with Brown Filter Cards
Disposable sample chambers with brown filter cards and caps   ·  Cell deposition area of 6mm in diameter (28mm2)  ·  Pre-attached brown filter card for slower absorbency  ·  Excellent for scanty specimens such as CSF  ·  
12 Available on GSA/VA Contract for Federal Government customers only. Iris Sample Processing CytoFuge* Filter Concentrators
For CytoFuge 2; Disposable; Vol.: 50 to 400µL; Box of 200 Filter Cards, 192 Concentrators
13 Thermo Scientific* Shandon* EZ Single Cytofunnel* Bench Stand
Bench Stand
14 Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific* Shandon* TPX Single Sample Chamber, Caps, and Filter Cards
For use with disposable white filter cards for sample volumes up to 0.5mL and for use with disposable brown filter cards for sample volumes of 0.4mL or less TPX Single Sample Chamber   ·  Reusable  ·  Autoclavable  ·   Filter
15 Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific* Shandon* EZ Double Cytofunnel*
Allows two sample deposition areas on a single slide   ·  Double the number of sample presentations per run  ·  Funnel markings correspond to slide markings for simple identification  ·  Pre-attached filter card  ·  Ideal for use with immunochemistry
  1. 1