Thermo Scientific™ Nalgene™ Autoclavable Carboys
Description Catalog Number Available Packaging
Capacity: 2.5 gal. (10L)
02-960-20C
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific™ No.: 22500020
Each
Case of 6 Each
Capacity: 5.0 gal. (20L)
02-960-20A
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific™ No.: 22500050
Each
Case of 4 Each
Capacity: 13.0 gal. (50L)
02-960-20B
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific™ No.: 22500130
Case
Key Features:
Key Features:
  • Convenient shoulder handles allow easy carrying and pouring, even with gloved hands
  • Available in 10, 20, and 50L sizes for a variety of biopharmaceutical applications
  • Graduated to contain in 1 gal. and 5L increments for ease of use
  • Nalgene Top Works Fluid Transfer Systems (Cat. No. 2135) permits convenient liquid aseptic transfer
  • Autoclavable/Graduated/Leakproof†
Ordering Information:
  • Additional closure options are Nalgene Quick Filling/Venting Closures (Cat. No. 2158) or Nalgene Filling/Venting Closures (Cat. No. 2162)
  • Additional accessories include Nalgene Barbed Bulk Head Fittings (Cat. No. 6149)
  • Custom Tubing Sets are available-please refer to Thermo Scientific Custom Tubing Set Configurator at www.thermoscientific.com/tubingconfigurator.com
  • 8210 Series is manufactured in our state-of-the-art Suzhou, China facility and is available for purchase in Asia Pacific region.  All other products manufactured in Rochester, NY.
Specifications
Autoclavable Yes
Closure Size 83B
Handles Yes
Material Polypropylene
No. per Pack 1