Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Diazabicyclo[5.4.0]undec-7-ene, 98+%, Thermo Scientific Chemicals

Catalog Number aaa1244914
Click to view available options
Quantity:
25 g
100 g
500 g
6674-22-2
C9H16N2
152.241
MFCD00006930
GQHTUMJGOHRCHB-UHFFFAOYSA-N
1,8-diazabicyclo 5.4.0 undec-7-ene, 2,3,4,6,7,8,9,10-octahydropyrimido 1,2-a azepine, polycat dbu, dbu, unii-h1ilj6ibux, pyrimido 1,2-a azepine, 2,3,4,6,7,8,9,10-octahydro, 1,8-diazabicyclo 5.4.0-7-undecene, h1ilj6ibux, 1,8-diazabicyclo 5,4,0 undec-7-ene
81184
2,3,4,6,7,8,9,10-octahydropyrimido[1,2-a]azepine
C1CCC2=NCCCN2CC1
6674-22-2
C9H16N2
152.241
MFCD00006930
GQHTUMJGOHRCHB-UHFFFAOYSA-N
1,8-diazabicyclo 5.4.0 undec-7-ene, 2,3,4,6,7,8,9,10-octahydropyrimido 1,2-a azepine, polycat dbu, dbu, unii-h1ilj6ibux, pyrimido 1,2-a azepine, 2,3,4,6,7,8,9,10-octahydro, 1,8-diazabicyclo 5.4.0-7-undecene, h1ilj6ibux, 1,8-diazabicyclo 5,4,0 undec-7-ene
81184
2,3,4,6,7,8,9,10-octahydropyrimido[1,2-a]azepine
C1CCC2=NCCCN2CC1

1,8-Diazabicyclo[5.4.0]undec-7-ene is used in organic synthesis as a catalyst, a complexing ligand, and a non-nucleophilic base. It is used as a protecting agent for the synthesis of cephalosporin and as a catalyst for polyurethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,8-Diazabicyclo[5.4.0]undec-7-ene is used in organic synthesis as a catalyst, a complexing ligand, and a non-nucleophilic base. It is used as a protecting agent for the synthesis of cephalosporin and as a catalyst for polyurethane.

Solubility
Miscible with water.

Notes
Incompatible with acids, strong oxidizing agents, acid anhydrides, acid chlorides and chloroformates.
TRUSTED_SUSTAINABILITY
Melting Point -70°C
Density 1.02
pH 12.8
Boiling Point 261°C
Flash Point 116°C (240°F)
Odor Amine-like
Refractive Index 1.522
Quantity 25 g
UN Number UN3267
Beilstein 508906
Sensitivity Air Sensitive
Solubility Information Miscible with water.
Formula Weight 152.24
Percent Purity 99%
Chemical Name or Material 1,8-Diazabicyclo[5.4.0]undec-7-ene
Show More Show Less
Hazard Category H301-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
EINECSNumber 229-713-7
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit