Learn More
101 BIO 1-Drop PCR Master Mix (with dye)
Supplier: 101 BIO W25995
User can squeeze one drop of PCR Master mix into PCR tube/plate, add template and primers, and then can run PCR reaction. This PCR Master Mix contains Taq DNA Polymerase, PCR buffer, Mg2+, dNTP, PCR stabilizer and PCR enhancer. This unique Mix recipe makes the system very reliable, and tolerant to most of the PCR condition (concentration of the components). The Taq Polymerase in the PCR Master Mix is Hot Start polymerase, which activity is inhibited at ambient temperatures by the chemical modificaiton. This prevents the formation of misprimed products and primer dimers at ambient temperatures. So no need to prepare the reactions on ice. The Polymerase is activated by a 10 minutes, 95°C incubation.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.