Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-1,3,2-dioxaphospholane-2-oxide, 95%, Thermo Scientific Chemicals

Catalog Number p-3735962
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
6609-64-9
C2H4ClO3P
142.48
MFCD00043138
SBMUNILHNJLMBF-UHFFFAOYSA-N
2-chloro-2-oxo-1,3,2-dioxaphospholane, ethylene chlorophosphate, 2-chloro-1,3,2-dioxaphospholane-2-oxide, 2-chloro-1,3,2-dioxaphospholane 2-oxide, ethylene glycol chlorophosphate, soqdebhb^bsvusup@, 2-chloro-1,3,2??-dioxaphospholan-2-one, 2-chlor-1,3,2-dioxaphospholan-2-oxid, acmc-1b2u0, ethylene cycl-chlorophosphate
81087
2-chloro-1,3,2$l^{5}-dioxaphospholane 2-oxide
C1COP(=O)(O1)Cl
6609-64-9
C2H4ClO3P
142.48
MFCD00043138
SBMUNILHNJLMBF-UHFFFAOYSA-N
2-chloro-2-oxo-1,3,2-dioxaphospholane, ethylene chlorophosphate, 2-chloro-1,3,2-dioxaphospholane-2-oxide, 2-chloro-1,3,2-dioxaphospholane 2-oxide, ethylene glycol chlorophosphate, soqdebhb^bsvusup@, 2-chloro-1,3,2??-dioxaphospholan-2-one, 2-chlor-1,3,2-dioxaphospholan-2-oxid, acmc-1b2u0, ethylene cycl-chlorophosphate
81087
2-chloro-1,3,2$l^{5}-dioxaphospholane 2-oxide
C1COP(=O)(O1)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 11.0°C to 14.0°C
Color White
Density 1.5500g/mL
Boiling Point 98.0°C to 99.0°C (2.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (ex Cl) (Argentometry)
Packaging Glass bottle
Refractive Index 1.4490 to 1.4540
Quantity 5 g
Specific Gravity 1.55
Formula Weight 142.48
Percent Purity 95%
Physical Form Solid Film
Chemical Name or Material 2-Chloro-1, 3, 2-dioxaphospholane-2-oxide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 229-560-6

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit