Learn More
Eisco™ 3 Prong Clamp with Boss head - Stainless Steel
Universal 3 pronged clamp with chemical resistant PVC coated jaws. 9.5 in. Total length with a convenient tightening screw for a secure hold. Clamp comes with boss head that opens to approximately 1 in.
Supplier: Eisco™ CH0684E
Specifications
Clamp | |
Metal |