Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromopyrazine-2-carboxylic acid, 95%, Thermo Scientific Chemicals

Catalog Number aah66382md
Click to view available options
Quantity:
250 mg
1 g
876161-05-6
C5H3BrN2O2
203.00
MFCD09909672
NPJHCKULJAIWGV-UHFFFAOYSA-N
18779790
5-bromopyrazine-2-carboxylic acid
OC(=O)C1=CN=C(Br)C=N1
876161-05-6
C5H3BrN2O2
203.00
MFCD09909672
NPJHCKULJAIWGV-UHFFFAOYSA-N
18779790
5-bromopyrazine-2-carboxylic acid
OC(=O)C1=CN=C(Br)C=N1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Synonym 5-bromo-2-pyrazinecarboxylic acid, 2-pyrazinecarboxylicacid, 5-bromo, 2-pyrazinecarboxylic acid, 5-bromo, 5-bromo-2-pyrazinecarboxylicacid, ksc446e0j, 2-pyrazinecarboxylicacid,5-bromo, 2-bromopyrazine-5-carboxylic acid
Formula Weight 203
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 5-Bromopyrazine-2-carboxylic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit