Learn More
Eppendorf™ Adapters for Rotors
Adapters for Rotors are for use with a variety of Eppendorf™, Corning™ and Nalgene™ tubes and plates in different shapes and sizes.
$422.50 - $464.00
Catalog Number | Mfr. No. | Capacity | Tube Type | For Use With (Equipment) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity | Tube Type | For Use With (Equipment) | Price | Quantity & Availability | |||
13-864-347
|
Eppendorf™
5920718002 |
2.6 to 8 mL | Round Bottom | Rotor S-4x1000, High-capacity Buckets |
Each for $427.50
|
|
|||
13-864-355
|
Eppendorf™
5920739000 |
2.6 to 8 mL | Round Bottom | Rotor S-4xUniversal-Large |
Each for $422.50
|
|
|||
13-864-358
|
Eppendorf™
5920742000 |
5 mL FACS | Round Bottom | Rotor S-4xUniversal-Large |
Each for $464.00
|
|
|||
13-864-354
|
Eppendorf™
5920738003 |
5.5 to 12 mL | Round Bottom | Rotor S-4xUniversal-Large |
Each for $422.50
|
|
|||
13-864-349
|
Eppendorf™
5920722000 |
9 mL | Round Bottom | Rotor S-4x1000, High-capacity Buckets |
Each for $427.50
|
|
|||
13-864-356
|
Eppendorf™
5920740008 |
250 mL Flat, 175 to 225 mL Conical | Flat/Conical Bottom | Rotor S-4xUniversal-Large |
Each for $463.00
|
|
|||
13-864-346
|
Eppendorf™
5920717006 |
250 mL Flat, 175 to 225 mL Conical, Nunc™ 250 mL Conical | Flat/Conical Bottom | Rotor S-4x1000, High-capacity Buckets |
Each for $454.50
|
|
|||
13-864-359
|
Eppendorf™
5920744003 |
500 mL Corning™ Conical Bottle | Conical | Rotor S-4xUniversal-Large |
Each for $463.00
|
|
|||
13-864-360
|
Eppendorf™
5920745000 |
500 mL Nalgene™ Flat Bottle | Flat Bottom | Rotor S-4xUniversal-Large |
Each for $463.00
|
|
|||
13-864-357
|
Eppendorf™
5920741004 |
750 mL Bottle | Flat Bottom | Rotor S-4xUniversal-Large |
Each for $463.00
|
|
|||
13-864-353
|
Eppendorf™
5920737007 |
6 x MTP, DWP, PCR Plates | Flat Bottom | S-4xUniversal-Large, Universal Buckets |
Each for $463.00
|
|
|||
Description
Available for use with:- 2.6 to 8 mL, set of 2 pcs.
- 250 mL flat, 175 to 225 mL conical, and Nunc™ 250 mL conical, set of 2 pcs.
- Please note that these conical bottles need to be used together with a conical form insert provided by the manufacturer.
- 49 x 2.6 to 8 mL, set of 2 pcs.
- 5.5 to 12 mL, set of 2 pcs.
- 500 mL Corning™, set of 2 pcs.
- 500 mL Nalgene™, set of 2 pcs.
- 50 mL, set of 2 pcs.
- 5 mL FACS, set of 2 pcs.
- 750 mL, set of 2 pcs.
- 9 mL, set of 2 pcs.
- Eppendorf Tubes™ 5.0 mL and 15 mL, set of 2 pcs.
- MTP, DWP, PCR plates, set of 2 pcs.
Specifications
Rotor Adapter |