Learn More
Thermo Scientific™ Air Filter Replacement Kits for Ultra Low Temperature Freezers
Easily replace the air filters on your ultra-low temperature freezer with these replacement parts.
Supplier: Thermo Scientific™ AF34567LE
Description
Easily replace the air filters on your ultra-low temperature freezer with Thermo Scientific™ Air Filter Replacement Kits for Ultra Low Temperature Freezers.
Replacement air filterWarranty and Services
Warranty varies by country, please contact us for details
Specifications
Replacement air filter | |
Freezer Air Filter | |
TSX Series, Revco RLE, Forma 89000, TLE Series, HERAfreeze HLE |