Learn More
AirClean™ Systems PowderSafe™ Type B Enclosure Accessory, Prefilter
SDP
AirClean™ Systems PowderSafe™ Type B Enclosure Accessory, Prefilter is an electrostatically charged fiber filter that removes sub-micron particulates.
Supplier:  AirClean™ Systems ACFPRE8
Catalog No. 36-100-0028
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • Electrostatically charged fiber filter that removes sub-micron particulates
  • Has efficiency rating of 99.997% at 0.3 microns
  • Used to protect absorption filters from contamination
  • Each absorption filter is fitted with a pre-filter at time of purchase
Specifications
Specifications
PowderSafe Type B Enclosure
Prefilter
Product Suggestions
Product Suggestions