Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Karl Fischer Aqualine™ Water Standard 1.0mg, for Karl Fischer Titration, Fisher Chemical

Catalog Number al271040
Click to view available options
Quantity:
10 x 4 mL
Packaging:
Ampoule
7732-18-5, 105-13-5, 100-66-3
240423
7732-18-5, 105-13-5, 100-66-3
240423

1mg/mL H2O standard

  • Standard contains 1mg H2O/mL (1000ppm)
  • Used for titer determination of coulometric or volumetric Karl Fischer reagents
  • Supplied as 10 x 4mL and packaged in glass ampules to protect from moisture
  • The ampule should only be opened immediately prior to use to prevent absorption of moisture from atmosphere
TRUSTED_SUSTAINABILITY
Color Colorless
Quantity 10 x 4 mL
pH 7
Equivalence 0.97 to 1.03mg of water/mL
Packaging Ampoule
Physical Form Liquid
Chemical Name or Material Aqualine™ Standard 1.1, Karl Fischer Reagent
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit