Learn More
Thermo Scientific™ Arctic Express™ Dual Storage Systems
Store and ship samples with Arctic Express dual storage systems, which feature hard-shell shipping containers.
$3530.00 - $3940.00
Specifications
Type Manual Fill LN2 Dewar
No. of Doors 1
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
11-675-900
Accessories View Documents
Thermo Scientific™
CK50920
10 L
Each for $3,530.00
Add to Cart
13-262-844
Accessories View Documents
Thermo Scientific™
CK50921
18.5 L
Each for $3,940.00
Add to Cart
Description
Description
Protect nonhazardous samples during shipment and storage using Thermo Scientific™ Arctic Express™ Storage Systems. Featuring excellent flexibility and safety features, these systems can be used as dry shippers or laboratory cryo vessels.
Features:
  • When filled with liquid nitrogen and placed in the laboratory, unit can be used as a “can and cane” storage system indefinitely with static holding times of up to 125 days.
  • Aluminum vessels come with lockable lid and hold six numbered, color-coded, stainless-steel cans (included), which accommodate canes that hold ampules.
  • Warranty varies by country, please contact us for details
Specifications
Specifications
Manual Fill LN2 Dewar
1