Learn More
Axygen™ 24- and 32-Well PCR Microplates, Clear, Nonsterile
GREENER_CHOICE
Designed to fit a wide variety of thermal cyclers
$214.00 - $301.00
Specifications
Certifications/Compliance RNase, DNase-free
Lid Without Lid
Color Clear
Product Type PCR Microplate, without Lid
Material Polypropylene
View More Specs
Products 2
Catalog Number Mfr. No. No. of Wells Price Quantity & Availability  
Catalog Number Mfr. No. No. of Wells Price Quantity & Availability  
14-222-301
GREENER_CHOICE
View Documents
Axygen™
PCR24C
24
Case of 50 for $214.00
Add to Cart
14-222-303
View Documents
Axygen™
PCR32C
32
Case of 50 for $301.00
Add to Cart
Description
Description
  • Volume to brim of well: 350 μL, working volume: 300 μL, volume with AxyMat: 240 μL
Specifications
Specifications
RNase, DNase-free
Clear
Polypropylene
Round
50/Cs.
Without Lid
PCR Microplate, without Lid
Non-sterile
PCR
10/Pk.