Learn More
Axygen™ Caps for PCR Tubes
8-Strip domed or flat strip caps for Axygen PCR tubes
$45.25 - $578.00
Catalog Number | Mfr. No. | Closure Type | Product Type | Color | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Closure Type | Product Type | Color | Price | Quantity & Availability | |||
14-222-265
|
Axygen™
PCR02CPC |
Domed | 8-Strip Domed PCR Tube Cap | Clear |
Pack of 125 for $60.50
Case of 10 PK for $514.50
|
|
|||
14-222-263
|
Axygen™
PCR02CPA |
Domed | 8-Strip Domed PCR Tube Cap | Assorted |
Pack of 125 for $71.90
Case of 10 PK for $600.50
|
|
|||
14-222-281
|
Axygen™
PCR02FCPC |
Flat | 8-Strip Flat PCR Tube Cap with Writing Surface | MaxyClear, w/writing surface |
Case of 1250 for $578.00
|
|
|||
14-222-302
|
Axygen™
PCR2CPRTC |
Flat | 8-Strip Flat PCR Tube Cap | MaxyClear |
Pack of 125 for $98.60
Case of 10 PK for $823.50
|
|
|||
14-223-222
|
Axygen™
PCR02CPC100 |
Domed | PCR Caps | Clear |
Case of 100 for $45.25
|
|
|||
Description
Axygen polypropylene, thin wall PCR tubes with and without caps are designed to fit standard 96-well blocks and are available in an assortment of colors. The consistent and ultra-thin wall allows precise thermal transfer. An even insertion cap ensures a tight fit to reduce evaporation.
Specifications
0.2 mL | |
Non-sterile | |
Polypropylene |