Learn More
beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt, 95%, Thermo Scientific Chemicals
Description
NADPH tetra sodium salt is used as an ubiquitous cofactor and biological reducing agent. β-NADPH is a coenzyme found in all living cells and is involved in redox reactions carrying electrons from one reaction to another. It is used as an electron donor, cofactor for many redox enzymes including nitric oxide synthetase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
NADPH tetra sodium salt is used as an ubiquitous cofactor and biological reducing agent. β-NADPH is a coenzyme found in all living cells and is involved in redox reactions carrying electrons from one reaction to another. It is used as an electron donor, cofactor for many redox enzymes including nitric oxide synthetase.
Solubility
Soluble in water (50 mg/ml).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C. Light sensitive. Keep away from strong oxidizing agents.
Specifications
Color | White |
Quantity | 100 mg |
Sensitivity | Light sensitive |
Solubility Information | Soluble in water (50mg/ml). |
Formula Weight | 833.35 |
Percent Purity | 95% |
Physical Form | Powder |
Chemical Name or Material | beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt |
Safety and Handling
EINECSNumber | 220-163-3 |
TSCA | No |
Recommended Storage | Store at -20°C |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.