Learn More
Bio Plas Siliconized Screw Cap Microcentrifuge Tubes
Ideal for packaging, transporting and storage of any diagnostic tests, especially in the area of biotechnology, biological products and reagents.
Specifications
Max. RCF | 25,000 x g |
---|---|
Color | Natural |
Material | Polypropylene |
Alert:
Siliconized products should NOT be autoclaved.
Catalog Number | Mfr. No. | Capacity (Metric) | Sterility | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Sterility | Price | Quantity & Availability | ||||
05-541-62
|
Bio Plas
4202SL |
1.5 mL | Non-sterile | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
05-541-61
|
Bio Plas
4200SLS |
0.5 mL | Sterile |
Item Discontinued
This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. View alternate product |
N/A | ||||
05-541-63
|
Bio Plas
4202SLS |
1.5 mL | Sterile |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | ||||
Description
Ideal for packaging, transporting and storage of any diagnostic tests, especially in the area of biotechnology, biological products and reagents.
Extremely low surface adhesion, which is ideal for protein or nucleic acid studies to prevent any possible loss of sample due to adherence to the inner walls of the tube allowing complete sample recovery.
- Positive screw cap seal with an O-ring designed to provide a safe, reliable secure fit even for repeated cap closures
- Can withstand centrifugal forces up to 25,000xG
- Conical, polypropylene, natural, available sterile or nonsterile
- A tight seal closure eliminates accidental opening during incubation, spinning or transporting
Order Info
Caps are sold separately (Fisher Cat No. 05-541-70
Specifications
25,000 x g | |
Natural | |
Polypropylene |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.