Learn More
Brady™ Code Reader™ 2600 Barcode Readers
Reads all 1D and 2D barcode symbologies
Catalog Number | Mfr. No. | Type | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Includes | Price | Quantity & Availability | ||||
19-104-469
|
Brady™
133080 |
Barcode Reader Handle Kit | Handle, bluetooth, battery, charging station with US power supply, and M3 modem with USB comm cable | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
19-104-470
|
Brady™
133081 |
Barcode Reader Handle Kit | Handle, bluetooth, and battery | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
19-104-471
|
Brady™
133082 |
Barcode Reader Palm Kit | Palm with bluetooth, battery, charging station with US power supply, and M3 modem with USB comm cable | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
19-104-472
|
Brady™
133083 |
Barcode Reader Palm Kit | Palm with Bluetooth and battery | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
Reads all 1D and 2D barcode symbologies
- Easy to use, lightweight, and high performing
- Features durable, rechargeable battery plus programmable buttons for custom workflow
- Battery status LED indicators with fuel gauge
- User feedback with vibration, audible tones and LED
- Patented dual field optics, both high density and wide field in same unit
- High speed, omnidirectional reading of 1D and 2D barcodes
- Unequaled performance on rounded, curved and shiny surfaces
- Multiple programmable buttons for customized work flow processes
- Easy to clean, disinfectant ready, IP65 housing for healthcare use
- Powerful data management capability with JavaScript
- Available in Handle and Palm kits