Learn More
BrandTech™ Support Jacks
Available in anodized aluminum (blue) or epoxy-powder-coated aluminum (green) decks
$257.50 - $2410.00
Specifications
Product Type | Support Jack |
---|---|
For Use With (Application) | Fume hoods and marine and industrial laboratories, POM rollers |
Catalog Number | Mfr. No. | Material | Height (English) | Length (English) Base | Width (English) Base | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Height (English) | Length (English) Base | Width (English) Base | Price | Quantity & Availability | ||
03-840-141
|
BrandTech™
B11021 |
Anodized Aluminum | 2.36 to 10.82 in. | 6.29 in. | 5.11 in. |
Each for $563.50
|
|
||
03-840-142
|
BrandTech™
B11031 |
Anodized Aluminum | 2.36 to 10.82 in. | 7.87 in. | 7.87 in. |
Each for $651.50
|
|
||
03-840-143
|
BrandTech™
B11041 |
Anodized Aluminum | 2.36 to 10.82 in. | 9.44 in. | 9.44 in. |
Each for $384.00
|
|
||
03-840-144
|
BrandTech™
B11081 |
Anodized Aluminum | 3.54 to 18.5 in. | 11.8 in. | 11.8 in. |
Each for $1,961.00
|
|
||
03-840-146
|
BrandTech™
B11015 |
Epoxy-powder Coated Aluminum | 2.16 to 4.72 in. | 3.93 in. | 3.93 in. |
Each for $257.50
|
|
||
03-840-147
|
BrandTech™
B11020 |
Epoxy-powder Coated Aluminum | 2.36 to 10.82 in. | 6.29 in. | 5.11 in. |
Each for $321.50
|
|
||
03-840-140
|
BrandTech™
B11016 |
Anodized Aluminum | 2.16 to 4.72 in. | 3.93 in. | 3.93 in. |
Each for $447.50
|
|
||
03-840-148
|
BrandTech™
B11030 |
Epoxy-powder Coated Aluminum | 2.36 to 10.82 in. | 7.87 in. | 7.87 in. |
Each for $351.00
|
|
||
03-840-149
|
BrandTech™
B11040 |
Epoxy-powder Coated Aluminum | 2.36 to 10.82 in. | 9.44 in. | 9.44 in. |
Each for $426.50
|
|
||
03-840-151
|
BrandTech™
B11090 |
Epoxy-powder Coated Aluminum | 3.5 to 18.5 in. | 15.7 in. | 15.7 in. |
Each for $1,904.00
|
|
||
03-840-145
|
BrandTech™
B11091 |
Anodized Aluminum | 3.5 to 18.5 in. | 15.7 in. | 15.7 in. |
Each for $2,410.00
|
|
||
Description
- Lift mechanism is made of 304 stainless steel with POM rollers for long service life
Specifications
Support Jack | |
Fume hoods and marine and industrial laboratories, POM rollers |