Learn More
Buffer Solution, pH 7.00, Color-Coded Yellow (Certified), Fisher Chemical™
$71.10 - $404.65
Specifications
Absorbance | Pass Test |
---|---|
Chemical Name or Material | Buffer Solution, pH 7.00, Color-Coded Yellow |
CAS | 7778-77-0, 1310-73-2, 7732-18-5, 1934-21-0 |
Melting Point | 0°C |
Boiling Point | 100°C |
Catalog Number | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Quantity | Packaging | Price | Quantity & Availability | |||||
SB107-500
|
500 mL | Poly Bottle |
Each for $71.10
Case of 6 EA for $340.50
Order in Bulk
Request bulk or custom formats |
|
|||||
SB107-4
|
4 L | Poly Bottle |
Each
for $207.65
Save 15% Reg :
Case of 4 EA
for $657.65
Save 15% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
SB107-20
|
20 L | PolyPac™ |
Each
for $404.65
Save 22% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
Specifications
Pass Test | |
7778-77-0, 1310-73-2, 7732-18-5, 1934-21-0 | |
100°C | |
1.0g/cm³ | |
7.00 | |
Pass Test | |
1 | |
760mmHg at 20°C | |
Certified |
Buffer Solution, pH 7.00, Color-Coded Yellow | |
0°C | |
Yellow | |
Potassium Phosphate Monobasic/Sodium Hydroxide. pH/temperature conversions given on label. | |
Liquid | |
MFCD00134130 | |
Traceable to NIST. | |
Soluble in water |
Safety and Handling
CAUTION!
Emergency Overview
May cause eye, skin, and respiratory tract irritation. Handle in accordance with good industrial hygiene and safety practice. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. . .
NFPA
Health:1
Flammability:0
Instability:0
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.