Learn More
BD Clay Adams Buckets for 4-Place Fixed-Angle Rotor for Dynac and Dynac III Centrifuges
Supplier: BD 420999
Description
Stainless-steel bucket fits directly in 05-100-28 and -32 rotors to accept heavy-duty 40mL tubes and regular strength 50mL tubes.
- Maximum tube dimensions: 30 dia. × 123mm long
- Supplied with cushions
Order Info
Buckets are sold one per package; order proper quantity to equip rotor
Specifications
Dynac™ and Dynac™ III centrifuge | |
Stainless Steel |