Learn More
Corning™ Ultra-Low Attachment Flasks
Features a covalently bound hydrogel layer that minimizes cell attachment, protein absorption and cellular activation. Corning™ Ultra-Low Attachment Flasks have an ergonomic shape that reduces the number of corners, improves cell scraping and allows the use of a larger pipet.
Specifications
Closure Color | Orange |
---|---|
Closure Material | HDPE |
Material | Polystyrene |
For Use With (Application) | For pipetting or scraping |
Neck Style | Canted |
Catalog Number | Mfr. No. | Capacity (Metric) | Shape | Culture Area | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Shape | Culture Area | Price | Quantity & Availability | |||
07-200-876
|
Corning™
3815 |
7.5 mL | Rectangular | 25 cm2 |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | |||
07-200-875
|
Corning™
3814CONV |
22.5 mL | U-shaped | 75 cm2 |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | |||
Description
- 25 to 75 cm2 available growth area
- Manufactured from optically clear virgin polystyrene
- Ultra-Low Attachment surface features a covalently bound hydrogel layer that effectively inhibits cellular attachment and minimizes protein absorption, enzyme activation, and cellular activation
- Surface is non-cytotoxic, biologically inert, and nondegradable
- Lot numbers for ease in traceability
- 100% integrity tested
- Sterilized by gamma irradiation
- Nonpyrogenic
Order Info
Sterilized by gamma radiation; certified nonpyrogenic
Specifications
Orange | |
Polystyrene | |
Canted | |
Ultra-low Attachment | |
Sterile |
HDPE | |
For pipetting or scraping | |
24/Cs. | |
Yes | |
Vented |
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.