Learn More
Decon™ AquaPur™ ST Sterile, Purified USP-Grade Water
For diluting sterile disinfectants
Supplier: Decon™ 8904
Alert:
Not for parenteral use. Not for injection, irrigation or infusion.
Description
Ideal for laboratory, cleanroom and manufacturing-facility applications.
- Filtered to 0.2μm and double-bagged in a Class 100 cleanroom
- Irradiated to SAL 10-6
- Tested for sterility and endotoxin using current USP method
Order Info
Lot-specific document shipped with each case
Compliance
Meets USP and CAPS/NCCL Type I specifications