Learn More
Thermo Scientific™ DNAPac™ PA200 Oligonucleotide HPLC Columns
Enjoy strong anion exchange for high-resolution analysis and purification of synthetic oligonucleotides with Thermo Scientific™ DNAPac™ PA200 Oligonucleotide HPLC Columns.
$1193.00 - $13040.00
Specifications
Pore Size | Nonporous |
---|---|
Temperature | 85°C |
Stationary Phase | Quaternary Ammonium |
Product Line | DNAPac |
Column Type | Ion Exchange |
Catalog Number | Mfr. No. | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
DX063000
|
Thermo Scientific™
063000 |
8 μm | 4 mm | 250 mm |
Each for $2,450.00
|
|
|||
DX063421
|
Thermo Scientific™
063421 |
8 μm | 9 mm | 250 mm |
Each for $5,020.00
|
|
|||
DX063425
|
Thermo Scientific™
063425 |
8 μm | 2 mm | 250 mm |
Each for $2,450.00
|
|
|||
DX062998
|
Thermo Scientific™
062998 |
8 μm | 4 mm | 50 mm |
Each for $1,193.00
|
|
|||
03-254-135
|
Thermo Scientific™
063419 |
8 μm | 9 mm | 50 mm |
Each for $1,325.00
|
|
|||
DX063423
|
Thermo Scientific™
063423 |
8 μm | 2 mm | 50 mm |
Each for $1,193.00
|
|
|||
DX082510
|
Thermo Scientific™
082510 |
4 μm | 4.6 mm | 250 mm |
Each for $4,800.00
|
|
|||
DX082509
|
Thermo Scientific™
082509 |
4 μm | 4.6 mm | 150 mm |
Each for $4,290.00
|
|
|||
DX082508
|
Thermo Scientific™
082508 |
4 μm | 4.6 mm | 50 mm |
Each for $3,560.00
|
|
|||
DXSP6734
|
Thermo Scientific™
088781 |
8 μm | 22 mm | 250 mm |
Each for $13,040.00
|
|
|||
08-878-0
|
Thermo Scientific™
088780 |
8 μm | 22 mm | 50 mm |
Each for $4,010.00
|
|
|||
Description
Enjoy strong anion exchange for high-resolution analysis and purification of synthetic oligonucleotides with Thermo Scientific™ DNAPac™ PA200 Oligonucleotide HPLC Columns.
- Achieve n, n-1 resolution for oligonucleotides
- Resolve oligonucleotides with secondary structures
- Assay phosphorothioate purity
- Selectivity control with eluent pH, salt, and solvent
- Resolve RNA with aberrant (2' – 5') links from normal SSRNA
Specifications
Nonporous | |
Quaternary Ammonium | |
Ion Exchange | |
Bio-LC |
85°C | |
DNAPac | |
Ion Exchange |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.