Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Drierite, with indicator, 8 mesh, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2 kg
5 kg
Packaging:
Glass Bottle
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent, Indicator
Specifications
Color | Blue |
Physical Form | Granular |
Quantity | 2 kg |
Fieser | 01,107 |
Merck Index | 15, 1708 |
Packaging | Glass Bottle |
Solubility Information | SiO2 and insolubles: 0.58% |
R2O | 0.29% |
IUPAC Name | calcium;sulfate |
Formula Weight | 136.13 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement: May cause cancer by inhalation. Suspected of causing genetic defects. May damage fertility. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause an allergic skin reaction. Very toxic to aquatic life with long lasting effect |
Precautionary Statement | GHS P Statement: Do not breathe dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. Avoid release to the environment. Obtain special instructions before use. Use personal protective equipment as required. Do not handle until all safety precautions have been read and understood. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction