Learn More
MilliporeSigma™ Isopore™ Membrane Filters, 0.8 μm
Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.
$135.90 - $558.00
Specifications
Autoclavable | Not Autoclavable |
---|---|
Bubble Point | 8.5 psi |
Color | White |
Filter Type | Screen Filter |
Material | Polycarbonate |
Catalog Number | Mfr. No. | Diameter (Metric) Filter | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) Filter | Price | Quantity & Availability | |||||
ATTP02500
|
MilliporeSigma™
ATTP02500 |
25 mm |
Pack of 100 for $151.50
|
|
|||||
ATTP01300
|
MilliporeSigma™
ATTP01300 |
13 mm |
Pack of 100 for $135.90
|
|
|||||
ATTP04700
|
MilliporeSigma™
ATTP04700 |
47 mm |
Pack of 100 for $223.00
|
|
|||||
ATTP14250
|
MilliporeSigma™
ATTP14250 |
142 mm |
Pack of 50 for $558.00
|
|
|||||
Description
Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.
The Isopore™ membrane is a polycarbonate, track-etched screen filter recommended for all analyses in which the sample is viewed on the surface of the membrane. Isopore™ membrane offers distinct advantages for the analysis of airborne contaminants and other particles using optical or electron microscopy. The Isopore™ membrane is composed of polycarbonate film, which has a smooth, glass-like surface for clearer sample observation. The unique manufacturing process of the membrane ensures a precise pore diameter and a consistent pore size for accurate separation of samples by size. Matched-weight filters are not usually required because of low, constant tar and ash weights.Features & Benefits:
•Membrane structure retains particles on the surface, simplifying counting and analysis
•Isopore™ membranes do not stain, resulting in low background interference
•Non-hygroscopic, allowing for rapid drying and reduced sample analysis time
•Translucent material does not require clearing for transmitted light microscopy; also available in brown variety
Applications:
Air Monitoring, Epifluorescent Microscopy, Chemotaxis Assays
Specifications
Not Autoclavable | |
White | |
Polycarbonate | |
Non-sterile | |
Air Analysis; Cell Biology; Cell Culture; Bioassays; Cytology; Environmental Analysis; Epifluorescence. | |
Hydrophilic | |
Hydrophilic |
8.5 psi | |
Screen Filter | |
0.8 μm | |
Avg. 85 mL/min x cm2 (typical results (at 10 psi);125 mL/min x cm2/psi) | |
20.0 L/min/cm2 | |
Filter Membranes |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.