Learn More
Invitrogen™ FLoid™ Cell Imaging Station
Captures high-quality, three-color fluorescent cell images right at your benchtopx
Supplier: Invitrogen™ 4471136
Description
Designed to remove all of the inherent complexities of capturing and processing three color fluorescent cell images, the EVOS FLoid Cell Imaging Station allows even the most novice users to spend their precious time on analyzing data rather than worrying about the operation of a microscope. Its interface is so simple, even novice users can collect data in just a few clicks of the mouse.
- Benchtop device for imaging cells
- Intuitive user interface allows novices and experts to generate fluorescence cell images in snap
- Self-Contained: no moving parts provides an insurance policy for your expensive confocal scopes allowing day-to-day use
- Integrated Molecular Probes™ reagent selection guide with protocols helps you design and execute your cell imaging experiments
- Print and stick captured images right in your notebook with associated printer (Sold separately: Cat. No. 10-213-413)
- Instrument design blocks ambient light taking fluorescent cell imaging out of darkroom and onto bench
- Comes with FLoid Cell Imaging Station Software and 2-gig USB drive
- Mechanical glide stage; range of motion, 10mm in X and Y dimensions
- Dimensions: 13.9D × 15.9W × 21.1 in.H (35.3 × 40.4 × 53.6cm)
- Weight: 26 lb. (11.8kg)
- Operating Power: 100-240 VAC, 50-60 Hz
- Electrical input: 5 VDC, 4.15A
- Operating Temperature: 4-32°C
- Operating Humidity: <90% (noncondensing)
- Output Ports: 4 USB
- LCD monitor: 15 in. color; display resolution 1366 x 768 pixels
- Image resolution 1296 x 964 pixels; adjustable tilt
- Captured Images: 16-bit monochochrome TIFF, PNG, BMP and JPG
- Objective: 20X plan fluorite
- Numerical aperture: 0.45 NA
- Working distance (WD): 5.9mm (defined as tip of objective to bottom of a 1.2mm thick slide)
- Overall Magnification: 460X (optical); 1840X (with digital zoom)
- Display Resolution: 1366 × 768 pixels
- Image Resolution: 1296 × 964 pixels
- Contrast Methods: Fluorescence and transmitted light (relief phase)
- Four Color Channels: Relief phase, blue fluorescence, green fluorescence, red fluorescence
- Illumination: LED (50,000 hr. life), adjustable intensity
- Excitation: Blue channel: 390/40nm; Green: 482/18nm; Red: 586/15nm
- Emission: Blue channel: 446/33nm; Green: 532/59nm; Red: 646/68nm
- Stage to Condenser: 60mm
- Stage: Range of motion, 4mm in X and Y dimensions
- Camera: Sony 1.3MP 1/3 in. ICX445 EXview HAD CCD
Specifications
Cell Imaging System | |
1.3 Megapixels, Monochrome, HAD CCD, 1/3 in. | |
Floid™ Cell Imaging System | |
Slide(s), Cell Culture Dish, Cell Culture Plate (Multi-Well), Cell Culture Flask (T75), Petri Dish, Coverslip(s) | |
FITC, Texas Red, DAPI | |
460X (Optical), 1840X (Digital) | |
Image Resolution - 1296 x 964 pixels, Display Resolution - 1366 x 768 pixels | |
JPG Files, PNG Files, BMP Files, TIFF Files | |
16 in. (W) x 21 in. (H) x 14 in. (D) | |
Floid™ Cell Imaging Station. Store dry at room temperature. The Floid™ Printer is not included and must be purchased separately. |
|
50/60 Hz |
Floid | |
Cellular Imaging | |
Green, Red, Blue | |
Ex: 390/40 (Blue), 482/18 (Green), 586/15 (Red); Em:446/33 (Blue), 532/59 (Green), 646/68 (Red) | |
LED | |
Fixed, Plan Fluorite | |
Open-Stage Platform for Most Cell Vessels | |
Adjustable Tilt, 15 in. Color | |
12.7 kg | |
Room Temperature | |
100/240 VAC |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.