Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ F1-ClipTip™ Good Laboratory Pipetting (GLP) Kits

Enjoy added convenience, with a variety of packaged pipettes in a range of volumes, compatible pipette tips and a pipette stand to support good laboratory pipetting in your lab.

$1023.65 - $2268.00

Specifications

Number of Channels 1
Type GLP Kit
View More Specs

Products 2
Catalog Number Mfr. No. Description Volume (Metric) Compatible Tips Price Quantity & Availability  
Catalog Number Mfr. No. Description Volume (Metric) Compatible Tips Price Quantity & Availability  
14-488-039N
View Documents
Thermo Scientific™
4701140N
F1-ClipTip GLP Kit 1: 1-10μL, 10-100 μL and 100-1000 μL 1 to 1000 μL ClipTip tips: 12.5 Ext, 200 and 1000
Each for $1,023.65

Save 22%

Reg : $1,318.00

The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Edge
Add to Cart
 
14-488-040N
View Documents
Thermo Scientific™
4701150N
F1-ClipTip GLP Kit 2: 2-20 μL, 20-200 μL, 100-1000 μL and 8-ch 30-300 μL 2 to 1000, 30 to 300 μL ClipTip tips: 20, 200, 300 and 1000
Each for $2,268.00
Add to Cart
 
Description

Description

Support good laboratory pipettingin your lab with the Thermo Scientific™ F1-ClipTip™ GLP Kits, which offer a variety of single channel pipettes or a mix of single channels and a multichannel pipette packaged with compatible ClipTip tips, and a convenient pipette stand. Select the kit that is right for you, and take advantage of the F1-ClipTip performance and lightweight design to maximize your comfort.

Worry-free pipetting is finally here with the only tip that clips.
Sealed in security of ClipTip Technology. Feel the difference.

Eliminate the frustration of banging tips on your pipette
With a light touch, each tip is clipped securely on the pipette.

Reduce the risk of loose or leaking tips

When ClipTip pipette tips are on, they are sealed and will not loosen. The ClipTip pipetting system remains sealed until you decide to eject tips, even in the most comprehensive applications, including multiple tip touches and mixing cycles.

Enable consistent and reproducible pipetting from user to user

Higher-quality results and more efficient research.

Learn more >

Ergonomically in-touch
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of RSI

Silent, effortless volume adjustment
Enjoy the silent and lighter volume adjustment mechanism with over 50% lower forces compared to earlier models.

Secure volume lock
The secure volume setting prevents undesirable volume drift during operation. Simply lift the plunger up to set volume and press down to lock in place with minimum effort.

Adjustable comfort
The 120° adjustable finger rest – for both right- and left-handed operator comfort – allows the hand to relax during pipetting cycles.

Super blow-out
150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50μL models and below.

Wide Selection of ClipTip Pipette Tips
Choose from seven sizes in 96-format: 12.5 Ext, 20, 50, 200, 300, 300 Ext,1000μL

Lifetime warranty
With convenient online registration, and may vary by country.

Kit 1 includes:
- Three Thermo Scientific™ F1-ClipTip pipettes from 1 μL to 1000 μL
-1-10 μL
-10-100 μL
-100-1000 μL
-Grip Strip for each handle to improve grip
-ClipTip tips: 12.5 Ext: 1 x 96; 200: 1 x 96; 1000: 1 x 96
-F-stand
-Good Laboratory Pipetting Guide
-Reagent reservoir demo pack
 
Kit 2 includes:
-Four Thermo Scientific™ F1-ClipTip pipettes from 2 μL to 1000 μL
-2-20 μL
-20-200 μL
-100-1000 μL
-8-ch 30-300 μL
-Grip Strip for each handle to improve grip
-ClipTip tips: 20: 1 x 96; 200: 1 x 96; 300: 1 x 96; 1000: 1 x 96
-F-stand
-Good Laboratory Pipetting Guide
-Reagent reservoir demo pack


Learn more about the complete ClipTip family >
Specifications

Specifications

1
GLP Kit
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit