Learn More
Thermo Scientific™ Glycogen, molecular biology grade
Increase the recovery of nucleic acids during preciptation with this inert carrier.
Supplier: Thermo Scientific™ R0561
Description
Glycogen is a highly purified polysaccharide derived from oysters. It is an inert carrier which significantly increases the recovery of nucleic acids by alcohol precipitation. Glycogen is insoluble in ethanol and forms a precipitate that traps target nucleic acids. During centrifugation, a visible pellet is formed, which greatly facilitates handling of the precipitated nucleic acids. Glycogen quantitatively precipitates nucleic acids from diluted solutions with a higher efficiency than that of tRNA, linear polyacrylamide, or sonicated DNA (see References 1-4).
We offer two formulations of aqueous glycogen solutions that allow for fast and efficient DNA or RNA recovery:
Glycogen, molecular biology grade, recommended for DNA precipitation (#R0561).
Glycogen, RNA grade, can be used for both RNA and DNA precipitation (#R0551).
Highlights
- Ideal for high recovery of oligonucleotides (> 8 bases) and low amounts of DNA/RNA (> 20 pg) from diluted solutions
- Forms clearly visible pellets
- Does not interfere with enzymatic reactions†
- Does not interfere with gel electrophoresis of nucleic acids
- Does not interfere with spectrophotometrical determination of DNA/RNA concentration, (A260 to 280 measurements)
- Compatible with DNA transformation and in vitro transfection
Specifications
Glycogen | |
Glycogen | |
2 x 0.25 mL | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.