Learn More
GSC Field Lines and Equipotential Lines
GSC Field Lines and Equipotential Lines allow users to plot electrical field and equipotential lines to gain a better visualization of the electromagnetic field created when electricity flows through an electrode.
Supplier: GSC ELEFLDLN
Description
Kit includes:
- 1 Trough with graph print
- 1 Aluminum needle
- 1 Rod with a socket
- 1 Multimeter
- 1 Cylindrical base
- 2 Bar electrodes
- 2 Disc electrodes
- 1 Ring electrode
- 1 Stand rod
- 1 Boss head
- 2 Sets of flexible plug leads
Instructions included
Specifications
Electricity Education | |
1 | |
95 oz. |
Multimeter, leads, trough, electrodes | |
15.5 in. |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov