Learn More
Fisherbrand™ accuFlame™ Natural Gas Burner
Stainless-steel grid top for better flame adjustment
Requires : 0.25 in. I.D. gas supply tubing recommended (not included)
Description
Stainless-steel grid top for better flame adjustment
- Provides better Meker Cone definition for easier fine tuning
- Distributes flame through 100 separate openings for uniform heating; flame arrester prevents flashback
- Provides twice the heat output (2930w/10,000 BTU/hr.) of straight-tube burners
- Features Venturi tube for thorough mixing of gas and air, and adjustable gas orifices and air shutter for complete combustion
- Burns natural gas at 10 cu. ft./hr.
- Rustproof chrome-plated brass body with no-tip, zinc-aluminum alloy base
Specifications
Natural Gas Burner | |
Brass, Stainless Steel, Zinc | |
3 cm | |
150 to 600 BTU | |
19 cm |
Natural Gas | |
1.18 in. | |
Venturi Tube for Thorough Mixing of Air and Gas and Adjustable Air Shutter and Gas Orifices for Complete Combustion | |
7.4 in. |