Learn More
Fisherbrand™ Zero Air Generator Accessory, Annual Maintenance Kit
For use with Zero Air Generator.
Catalog No. 14-950-655
Add to Cart
Includes: Replacement filters
Description
Description
Helps protect catalyst module and downstream applications form contamination
Kit Components:
  • Prefilter
  • Sintered metal filter
Specifications
Specifications
Annual filter kit for 14950653 or 14950654
Zero Air Generators (14950654 and 14950655)
Prefilter and sintered metal filter
Maintenance Kit
Gas chromatography, flame support gas, flame ionization detector (GC-FID) flame photometric detector (GC-FPD)
Product Suggestions
Product Suggestions