Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Autoclavable HD Biohazard Waste Bags
Biohazard symbol printed bags to ensure safe handling and disposal of biohazardous waste.
$150.40 - $402.50
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | ASTM |
Closure Type | None |
Color | Orange with Black Print |
Includes | Printed in English |
Catalog Number | Dimensions (L x W) | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Dimensions (L x W) | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
01-815A
|
24 x 12 in. (61 x 30 cm) | 61 cm | 30 cm |
Case of 100 for $150.40
|
|
||||
01-815C
|
35 x 24 in. (88.9 x 61 cm) | 88.9 cm | 61 cm |
Case of 100 for $402.50
|
|
||||
01-815B
|
30 x 24 in. (76 x 61 cm) | 76 cm | 61 cm |
Case of 100 for $287.50
|
|
||||
Description
Impact- and tear-resistant autoclave bags for waste sterilization. Fisherbrand™ Orange Autoclave Bags without Sterilization Indicator are constructed of high-density polyethylene.
- Impact- and tear-resistant.
- Bright orange color.
- High-density polyethylene construction
Compliance
Pass ASTM™ impact- and tear-resistance tests.
Specifications
Autoclavable | |
None | |
Printed in English | |
38 micron | |
127°C |
ASTM | |
Orange with Black Print | |
HDPE | |
260°F |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title