Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ High Precision Dissecting Micro Scissors
SDP

Fisherbrand Dissecting Micro Scissors are powered by an automatic, spring-action mechanism which allow for ease and comfort of use for high precision applications.

Catalog No. 08-953-1B


Add to Cart

Description

Description

  • Fisherbrand™ Dissecting Micro Scissors are powered by a spring-action mechanism which allow for ease and comfort of use for high precision applications. Our insturments are designed to exceed performance expectations -This is the Fisherbrand Advantage. Available in straight or curved tip-style.
  • Squeeze gently for scissor action
  • Micro Blades; Sharp Tips; 4.75 in.
  • Multipurpose - Useful in educational, research or healthcare setting
  • Made from surgical grade stainless steel
  • Electronically passivated for corrosion resistance
  • Rockwell hardness (HRC): 46-50
  • Latex-free
  • Can be sterilized by Eto/Gamma radiation; Fully Autoclavable
  • Quality control per U.S. Government specs
  • Produced under ISO 9002, GMP, cGMP guidelines
Specifications

Specifications

Surgical Scissors
4.75 in.
Stainless Steel
Surgical, Precision
Straight
12 cm
Autoclavable
Sharp/Sharp
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit