Learn More
Thermo Scientific™ HyperSep™ SLE Cartridges (pH 9)
A fast and effective sample preparation technique for removal of phospholipids from biological matricies.
$163.80 - $204.50
Specifications
For Use With (Application) Removal of Phospholipids From Biological Matrices
Product Line HyperSep
Sorbent HyperSep Special Treated Diatomite
Type Solid Phase Extraction Cartridge
View More Specs
Products 3
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
03-251-394
View Documents
Thermo Scientific™
601092000129
2 g/12 mL, Pk. of 20, pH9
Pack of 20 for $167.20
Add to Cart
60-109-100069
View Documents
Thermo Scientific™
60109100069
1 g/6 mL, Pk. of 30, pH9
Pack of 30 for $163.80
Add to Cart
03-251-395
View Documents
Thermo Scientific™
601094000259
4 g/25 mL, Pk. of 15, pH9
Pack of 9 for $204.50
Add to Cart
Description
Description
Provide a fast and effective sample preparation technique for removal of phospholipids from biological matrices with Thermo Scientific™ HyperSep™ SLE (Solid supported Liquid/Liquid Extraction).
HyperSep SLE cartrides (pH9) deals with sample preparation of biological matrices via a simple, efficient and an effective protcol. The focus being the removal of interfering phospholipids which cause a number of issues for end point analysis.
SLE offers a number of advantages over existing sample preparation techniques such as Liquid/Liquid Extraction (LLE) and Protein Precipitation techniques for dealing with biological matrices. In comparison to LLE, SLE offers greater reproducibility and recoveries, lower solvent requirements and prevents emulsification which is often associated with the LLE technique. It also lends itself to full automation which is not possible via LLE. SLE provides improved cleanliness of sample extract and associated increase in sensitivity compared to protein precipitation techniques which are unable to remove phospholipids.
  • Simple, easy-to-use protocol
  • Removal of phospholipids
  • Greater reproducability and recoveries compared to LLE
Specifications
Specifications
Removal of Phospholipids From Biological Matrices
HyperSep
HyperSep Special Treated Diatomite
Solid Phase Extraction Cartridge