Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ JTV 519 fumarate
Ryanodine receptor (RyR) inhibitor
$247.08 - $1032.40
Specifications
Purity | 98% |
---|---|
Content And Storage | Store at +4°C |
Description | JTV 519 fumarate |
Gene Alias | K 201 |
Molecular Weight (g/mol) | M.W. (Observed): 540.67 |
Description
Ryanodine receptor (RyR) inhibitor; stabilizes RyR2 in a closed conformation. Exhibits antiarrhythmic and cardioprotective properties in vivo.Specifications
98% | |
JTV 519 fumarate | |
M.W. (Observed): 540.67 |
Store at +4°C | |
K 201 | |
JTV 519 fumarate |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title