Learn More
Keener™ Autoclave Bag Elastic Closures
An alternative to wire tie closures
Supplier: Keener™ 4 4X1 4HR
Description
An alternative to wire tie closures
- High-tensile-strength elastic bands
- Compatible with most autoclave bags
- Remain intact at autoclaving temperatures
Specifications
Autoclavable | |
Elastic |