Learn More
L-Proline, 99%, Thermo Scientific Chemicals
Description
L-Proline is used as asymmetric catalysts in organic synthesis and asymmetric aldol cyclization. It is involved in the Michael addition of dimethyl malonate to alfa-beta-unsaturated aldehydes. It is a precursor of hydroxyproline in collagen. It is an active component of collagen and involved in the proper functioning of joints and tendons. It finds uses in pharmaceutical, biotechnological applications due to its osmoprotectant property. Further, it is used with ninhydrin in the chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
L-Proline is used as asymmetric catalysts in organic synthesis and asymmetric aldol cyclization. It is involved in the Michael addition of dimethyl malonate to alfa-beta-unsaturated aldehydes. It is a precursor of hydroxyproline in collagen. It is an active component of collagen and involved in the proper functioning of joints and tendons. It finds uses in pharmaceutical, biotechnological applications due to its osmoprotectant property. Further, it is used with ninhydrin in the chromatography.
Solubility
Soluble in water and ethanol. Insoluble in ether, butanol and isopropanol.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications
Melting Point | ∽225°C (decomposition) |
Beilstein | 80810 |
Solubility Information | Soluble in water and ethanol. Insoluble in ether,butanol and isopropanol. |
Formula Weight | 115.13 |
Density | 1.36 |
Quantity | 25 g |
Sensitivity | Hygroscopic |
Chemical Name or Material | L-Proline |
Safety and Handling
EINECSNumber | 205-702-2 |
RTECSNumber | TW3584000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.