Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 2 ports, capsule filter and MPC insert

Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.

$1343.00 - $1437.00

Specifications

Product Type Bioprocess Container
Content And Storage Store at room temperature.
Includes 2 Ports (1 Membrane Capsule Filter and 1 MPC Insert With Pinch Clamps)
Material Polyethylene
Tubing Style C-Flex, 91.4 cm (36 in.)
View More Specs

Products 3
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
SS00162I
View Documents
Thermo Scientific™
SS00162I
50 L
Case of 5 for $1,361.00
Add to Cart
 
SS00163I
View Documents
Thermo Scientific™
SS00163I
100 L
Case of 5 for $1,343.00
Add to Cart
 
SS00164I
View Documents
Thermo Scientific™
SS00164I
200 L
Case of 5 for $1,437.00
Add to Cart
 
Description

Description

This Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL film. This BPC includes 2 ports (1 membrane capsule filter and 1 MPC insert with pinch clamps). It is available in 50, 100 and 200L capacities.

  • Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
  • All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications:

  • Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
  • Preparing and storing buffer, culture media, and process liquid
  • Collecting chromatography feed and fraction
  • Harvesting, storing, and transporting bulk drug product and drug precursors
Specifications

Specifications

Bioprocess Container
2 Ports (1 Membrane Capsule Filter and 1 MPC Insert With Pinch Clamps)
C-Flex, 91.4 cm (36 in.)
Two 4-mil. poly bags
Labtainer™
5/Cs.
Irradiated
Store at room temperature.
Polyethylene
ASI™ 26/77
Capsule filter and MPC insert + cap
Protective material around rough areas
Room Temperature
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit