Learn More
Eisco™ Linear Air Track with Accessories
SCI_ED
Study all aspects of dynamics in a virtually friction free environment with this cost effective system. The Linear Air Track contains everything required for a vast array of experiments.
Supplier:  Eisco™ PH0362A
Catalog No. S12954
Add to Cart
Description
Description
  • The Linear Air Track contains everything required for a vast array of experiments covering velocity, force and acceleration, potential energy, kinetic energy, conservation of energy, SHM, motion on level and inclined planes.
  • Cost effective system for the study of all aspects of dynamics.
  • Ideal for classroom use.
  • Virtually friction free for accurate demonstrations.
  • Designed to accept standard slotted masses.
  • Includes a range of accessories.
Specifications
Specifications
Linear Air Track
162.1 x 18 x 32 cm (63.8 x 7.1 x 12.6 in.)
32 cm
162.1 cm
18 cm
1.5 M Air Track (1) 120 G Slider with Mounting Bracket (2) Attachment Nuts (1) Spring Attachments (4) Connection Bracket (for Springs, String, etc) (4) 2 in. Spring (2) 4 in. Spring (2) Male Velcro Bumper (2) Female Velcro Bumper (2) Friction-less Pulley and Bracket (2) String (20 Ft.) 5 G Slotted Masses (9) Slotted Mass Hanger (1) Photogate Interruption Screens (8) Elastic Launching Cradle / Bumper (1) 12.5 G Slider Masses (4) 25 G Slider Masses (4) Photogate/accessory Attachment Arm (2) Leveling Feet (3) Air Input Valve (1) Measuring Meter on Track (1)
12.6 in.
63.8 in.
7.1 in.
24.9 lb.
Product Suggestions
Product Suggestions